Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67417590-67417829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARNS1
Entrez Gene ID
57571
Gene Name
carnosine synthase 1
Gene Aliases
ATPGD1
UniGene
Hs.502982
Gene Chromosome Location
Chr.11: 67415678-67425607 on Build GRCh38
Transcripts
NM_001166222.1 NM_020811.1 XM_011545189.2 XM_011545191.2 XM_017018054.1 XM_017018055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4951581 c.230C>A p.A77D Chr.11: 67417633-67417633 on Build GRCh38
6426460 c.269G>A p.R90H Chr.11: 67417672-67417672 on Build GRCh38
5574468 c.274G>A p.G92S Chr.11: 67417677-67417677 on Build GRCh38
6418101 c.254G>T p.R85L Chr.11: 67417657-67417657 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879750332 Chr.11: 67417671-67417671 on Build GRCh38
rs35295058 Chr.11: 67417750-67417751 on Build GRCh38
rs996079248 Chr.11: 67417698-67417698 on Build GRCh38
rs551197023 Chr.11: 67417785-67417785 on Build GRCh38
rs1048808244 Chr.11: 67417652-67417652 on Build GRCh38
rs80017078 Chr.11: 67417777-67417777 on Build GRCh38
rs911643821 Chr.11: 67417700-67417700 on Build GRCh38
rs559208092 Chr.11: 67417728-67417728 on Build GRCh38
rs973461598 Chr.11: 67417644-67417644 on Build GRCh38
rs990600351 Chr.11: 67417763-67417763 on Build GRCh38
rs976921932 Chr.11: 67417726-67417726 on Build GRCh38
rs1022883234 Chr.11: 67417621-67417621 on Build GRCh38
rs762448737 Chr.11: 67417756-67417756 on Build GRCh38
rs1028920565 Chr.11: 67417713-67417713 on Build GRCh38
rs760009624 Chr.11: 67417636-67417636 on Build GRCh38
rs764850754 Chr.11: 67417657-67417657 on Build GRCh38
rs528055975 Chr.11: 67417659-67417659 on Build GRCh38
rs944207283 Chr.11: 67417718-67417718 on Build GRCh38
rs183572582 Chr.11: 67417784-67417784 on Build GRCh38
rs913321556 Chr.11: 67417769-67417769 on Build GRCh38
rs528892595 Chr.11: 67417650-67417650 on Build GRCh38
rs561765457 Chr.11: 67417637-67417637 on Build GRCh38

View Full Product Details