Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31603605-31603834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP4
Entrez Gene ID
26610
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
UniGene
Hs.175534
Gene Chromosome Location
Chr.11: 31509729-31784525 on Build GRCh38
Transcripts
NM_001288725.1 NM_001288726.1 NM_019040.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5480858 c.524T>C p.V175A Chr.11: 31603778-31603778 on Build GRCh38
4032353 c.546C>T p.H182H Chr.11: 31603800-31603800 on Build GRCh38
926333 c.561A>G p.S187S Chr.11: 31603815-31603815 on Build GRCh38
4032351 c.527C>A p.S176* Chr.11: 31603781-31603781 on Build GRCh38
7119637 c.514-1G>C p.? Chr.11: 31603767-31603767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367822897 Chr.11: 31603779-31603779 on Build GRCh38
rs761129704 Chr.11: 31603716-31603716 on Build GRCh38
rs750554199 Chr.11: 31603749-31603749 on Build GRCh38
rs902793199 Chr.11: 31603678-31603678 on Build GRCh38
rs548106901 Chr.11: 31603796-31603796 on Build GRCh38
rs559717548 Chr.11: 31603744-31603744 on Build GRCh38
rs566381038 Chr.11: 31603805-31603805 on Build GRCh38
rs564056030 Chr.11: 31603681-31603681 on Build GRCh38
rs1004358337 Chr.11: 31603639-31603639 on Build GRCh38
rs771600237 Chr.11: 31603802-31603802 on Build GRCh38
rs769800146 Chr.11: 31603764-31603764 on Build GRCh38
rs754092897 Chr.11: 31603722-31603722 on Build GRCh38
rs1012295160 Chr.11: 31603739-31603739 on Build GRCh38
rs748105879 Chr.11: 31603754-31603754 on Build GRCh38
rs552329556 Chr.11: 31603673-31603673 on Build GRCh38
rs374800267 Chr.11: 31603767-31603767 on Build GRCh38
rs956106209 Chr.11: 31603747-31603748 on Build GRCh38
rs764446036 Chr.11: 31603721-31603721 on Build GRCh38
rs774913398 Chr.11: 31603803-31603803 on Build GRCh38
rs963233030 Chr.11: 31603666-31603666 on Build GRCh38
rs749033348 Chr.11: 31603771-31603771 on Build GRCh38
rs758527327 Chr.11: 31603752-31603752 on Build GRCh38
rs778994498 Chr.11: 31603743-31603743 on Build GRCh38
rs1021187711 Chr.11: 31603675-31603675 on Build GRCh38
rs771547884 Chr.11: 31603751-31603751 on Build GRCh38
rs570234056 Chr.11: 31603808-31603808 on Build GRCh38
rs35843493 Chr.11: 31603754-31603754 on Build GRCh38
rs867853166 Chr.11: 31603651-31603651 on Build GRCh38
rs780244331 Chr.11: 31603753-31603753 on Build GRCh38
rs113985892 Chr.11: 31603715-31603715 on Build GRCh38
rs745638917 Chr.11: 31603650-31603650 on Build GRCh38
rs773001512 Chr.11: 31603780-31603780 on Build GRCh38
rs757553138 Chr.11: 31603729-31603729 on Build GRCh38

View Full Product Details