Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136677847-136678120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF6
Entrez Gene ID
9459
Gene Name
Rac/Cdc42 guanine nucleotide exchange factor 6
Gene Aliases
COOL2, Cool-2, MRX46, PIXA, alpha-PIX, alphaPIX
UniGene
Hs.522795
Gene Chromosome Location
Chr.X: 136665547-136781656 on Build GRCh38
Transcripts
NM_001306177.1 NM_004840.2 XM_005262499.2 XM_011531412.2 XM_011531413.1 XM_011531414.1 XM_011531415.2 XM_011531416.2 XM_011531417.1 XM_017029975.1 XM_017029976.1

SNP Detail

SNP ID Chromosome Location
rs768648280 Chr.X: 136677945-136677945 on Build GRCh38
rs771926399 Chr.X: 136677977-136677977 on Build GRCh38
rs910450911 Chr.X: 136678085-136678085 on Build GRCh38
rs145014969 Chr.X: 136678003-136678003 on Build GRCh38
rs750917351 Chr.X: 136677950-136677950 on Build GRCh38
rs771016461 Chr.X: 136677911-136677911 on Build GRCh38
rs765922194 Chr.X: 136677990-136677990 on Build GRCh38
rs771615230 Chr.X: 136677980-136677980 on Build GRCh38
rs190459455 Chr.X: 136678016-136678016 on Build GRCh38
rs979485207 Chr.X: 136678012-136678012 on Build GRCh38
rs368720089 Chr.X: 136677906-136677906 on Build GRCh38
rs992529877 Chr.X: 136677879-136677879 on Build GRCh38
rs760413082 Chr.X: 136677981-136677981 on Build GRCh38
rs774280200 Chr.X: 136677946-136677946 on Build GRCh38
rs139592017 Chr.X: 136677880-136677880 on Build GRCh38
rs372548244 Chr.X: 136677889-136677889 on Build GRCh38
rs369871910 Chr.X: 136677900-136677900 on Build GRCh38
rs957391039 Chr.X: 136678063-136678063 on Build GRCh38
rs745588480 Chr.X: 136677922-136677922 on Build GRCh38
rs781383556 Chr.X: 136677917-136677917 on Build GRCh38
rs761684009 Chr.X: 136677947-136677947 on Build GRCh38
rs959973375 Chr.X: 136677876-136677876 on Build GRCh38
rs925792056 Chr.X: 136678013-136678013 on Build GRCh38
rs147574171 Chr.X: 136678077-136678077 on Build GRCh38

View Full Product Details