Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141929547-141929816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0141
Entrez Gene ID
9812
Gene Name
KIAA0141
Gene Aliases
DELE
UniGene
Hs.210532
Gene Chromosome Location
Chr.5: 141923785-141942047 on Build GRCh38
Transcripts
NM_001142603.2 NM_014773.4 XM_005268547.3 XM_005268548.3 XM_005268552.2 XM_011537715.1 XM_011537716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211839 c.515G>A p.R172Q Chr.5: 141929684-141929684 on Build GRCh38
736567 c.442G>A p.D148N Chr.5: 141929611-141929611 on Build GRCh38
7373425 c.418C>T p.R140* Chr.5: 141929587-141929587 on Build GRCh38
6692974 c.564T>A p.S188S Chr.5: 141929733-141929733 on Build GRCh38
4407079 c.531C>T p.N177N Chr.5: 141929700-141929700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761254815 Chr.5: 141929765-141929765 on Build GRCh38
rs76364748 Chr.5: 141929644-141929644 on Build GRCh38
rs759352073 Chr.5: 141929683-141929683 on Build GRCh38
rs371713234 Chr.5: 141929706-141929706 on Build GRCh38
rs758625390 Chr.5: 141929708-141929708 on Build GRCh38
rs530284805 Chr.5: 141929635-141929635 on Build GRCh38
rs746200012 Chr.5: 141929626-141929626 on Build GRCh38
rs756799409 Chr.5: 141929702-141929702 on Build GRCh38
rs114623503 Chr.5: 141929587-141929587 on Build GRCh38
rs201802656 Chr.5: 141929611-141929611 on Build GRCh38
rs114907095 Chr.5: 141929595-141929595 on Build GRCh38
rs770012506 Chr.5: 141929719-141929719 on Build GRCh38
rs376636019 Chr.5: 141929577-141929577 on Build GRCh38
rs10078802 Chr.5: 141929700-141929700 on Build GRCh38
rs372468560 Chr.5: 141929792-141929792 on Build GRCh38
rs138993288 Chr.5: 141929688-141929688 on Build GRCh38
rs781775516 Chr.5: 141929716-141929716 on Build GRCh38
rs781743807 Chr.5: 141929780-141929780 on Build GRCh38
rs374773989 Chr.5: 141929724-141929724 on Build GRCh38
rs775339536 Chr.5: 141929728-141929728 on Build GRCh38
rs201186658 Chr.5: 141929624-141929624 on Build GRCh38
rs755518956 Chr.5: 141929608-141929608 on Build GRCh38
rs764061758 Chr.5: 141929754-141929754 on Build GRCh38
rs748682491 Chr.5: 141929781-141929781 on Build GRCh38
rs6888914 Chr.5: 141929789-141929789 on Build GRCh38
rs369039562 Chr.5: 141929769-141929769 on Build GRCh38
rs751918075 Chr.5: 141929772-141929772 on Build GRCh38
rs141459129 Chr.5: 141929682-141929682 on Build GRCh38
rs144583494 Chr.5: 141929704-141929704 on Build GRCh38
rs1037568526 Chr.5: 141929761-141929761 on Build GRCh38
rs776762258 Chr.5: 141929638-141929638 on Build GRCh38
rs755491125 Chr.5: 141929714-141929714 on Build GRCh38
rs773506426 Chr.5: 141929720-141929720 on Build GRCh38
rs766866131 Chr.5: 141929730-141929731 on Build GRCh38
rs145156674 Chr.5: 141929684-141929684 on Build GRCh38
rs759448332 Chr.5: 141929596-141929596 on Build GRCh38
rs752698543 Chr.5: 141929607-141929607 on Build GRCh38
rs548064875 Chr.5: 141929625-141929625 on Build GRCh38
rs564159851 Chr.5: 141929775-141929775 on Build GRCh38
rs201111574 Chr.5: 141929699-141929699 on Build GRCh38
rs955898498 Chr.5: 141929756-141929756 on Build GRCh38
rs776651281 Chr.5: 141929588-141929588 on Build GRCh38
rs747392410 Chr.5: 141929713-141929713 on Build GRCh38
rs200808757 Chr.5: 141929768-141929768 on Build GRCh38
rs571217703 Chr.5: 141929739-141929739 on Build GRCh38
rs780464113 Chr.5: 141929711-141929711 on Build GRCh38
rs567971593 Chr.5: 141929634-141929634 on Build GRCh38
rs570183351 Chr.5: 141929689-141929689 on Build GRCh38
rs372768684 Chr.5: 141929783-141929783 on Build GRCh38
rs200197275 Chr.5: 141929686-141929686 on Build GRCh38
rs747556845 Chr.5: 141929636-141929636 on Build GRCh38
rs151086425 Chr.5: 141929651-141929651 on Build GRCh38
rs769111411 Chr.5: 141929637-141929637 on Build GRCh38
rs776581221 Chr.5: 141929764-141929764 on Build GRCh38
rs372423631 Chr.5: 141929567-141929567 on Build GRCh38
rs533546209 Chr.5: 141929766-141929766 on Build GRCh38
rs765616804 Chr.5: 141929590-141929590 on Build GRCh38
rs770113742 Chr.5: 141929650-141929650 on Build GRCh38
rs767313892 Chr.5: 141929599-141929599 on Build GRCh38
rs749526729 Chr.5: 141929722-141929722 on Build GRCh38
rs545198440 Chr.5: 141929589-141929589 on Build GRCh38
rs199887534 Chr.5: 141929579-141929579 on Build GRCh38
rs777999050 Chr.5: 141929788-141929788 on Build GRCh38
rs777339310 Chr.5: 141929610-141929610 on Build GRCh38
rs748202306 Chr.5: 141929717-141929717 on Build GRCh38
rs1007473228 Chr.5: 141929779-141929779 on Build GRCh38
rs558109011 Chr.5: 141929775-141929775 on Build GRCh38

View Full Product Details