Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32771074-32771329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1522
Entrez Gene ID
57648
Gene Name
KIAA1522
Gene Aliases
-
UniGene
Hs.591502
Gene Chromosome Location
Chr.1: 32741839-32774970 on Build GRCh38
Transcripts
NM_001198972.1 NM_001198973.1 NM_020888.2 XM_006710791.3 XM_017001916.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7147183 c.1810C>T p.H604Y Chr.1: 32771166-32771166 on Build GRCh38
7147337 c.2024C>G p.S675C Chr.1: 32771203-32771203 on Build GRCh38
5846413 c.2108G>A p.S703N Chr.1: 32771287-32771287 on Build GRCh38
123947 c.1913C>T p.S638F Chr.1: 32771092-32771092 on Build GRCh38
3865331 c.2012C>T p.S671F Chr.1: 32771191-32771191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748722328 Chr.1: 32771170-32771170 on Build GRCh38
rs747348119 Chr.1: 32771137-32771137 on Build GRCh38
rs767529564 Chr.1: 32771102-32771102 on Build GRCh38
rs780358008 Chr.1: 32771121-32771121 on Build GRCh38
rs371646422 Chr.1: 32771282-32771282 on Build GRCh38
rs759435830 Chr.1: 32771099-32771099 on Build GRCh38
rs758488873 Chr.1: 32771113-32771113 on Build GRCh38
rs760537844 Chr.1: 32771293-32771293 on Build GRCh38
rs117872916 Chr.1: 32771244-32771244 on Build GRCh38
rs771851259 Chr.1: 32771285-32771285 on Build GRCh38
rs766493218 Chr.1: 32771224-32771224 on Build GRCh38
rs752882300 Chr.1: 32771228-32771229 on Build GRCh38
rs763909500 Chr.1: 32771301-32771301 on Build GRCh38
rs541220282 Chr.1: 32771209-32771209 on Build GRCh38
rs772539926 Chr.1: 32771177-32771177 on Build GRCh38
rs990681861 Chr.1: 32771273-32771273 on Build GRCh38
rs559528999 Chr.1: 32771126-32771126 on Build GRCh38
rs1012522329 Chr.1: 32771131-32771131 on Build GRCh38
rs377142667 Chr.1: 32771107-32771107 on Build GRCh38
rs765297662 Chr.1: 32771109-32771109 on Build GRCh38
rs748852883 Chr.1: 32771187-32771187 on Build GRCh38
rs755387345 Chr.1: 32771147-32771147 on Build GRCh38
rs199566536 Chr.1: 32771223-32771223 on Build GRCh38
rs745578874 Chr.1: 32771283-32771283 on Build GRCh38
rs775561100 Chr.1: 32771216-32771216 on Build GRCh38
rs1027865789 Chr.1: 32771161-32771161 on Build GRCh38
rs201507589 Chr.1: 32771243-32771243 on Build GRCh38
rs767476662 Chr.1: 32771212-32771212 on Build GRCh38
rs759380910 Chr.1: 32771211-32771211 on Build GRCh38
rs764102402 Chr.1: 32771221-32771221 on Build GRCh38
rs369607424 Chr.1: 32771150-32771150 on Build GRCh38
rs750418650 Chr.1: 32771222-32771222 on Build GRCh38
rs760460932 Chr.1: 32771220-32771220 on Build GRCh38
rs376897230 Chr.1: 32771225-32771225 on Build GRCh38
rs988283348 Chr.1: 32771262-32771262 on Build GRCh38
rs1044039798 Chr.1: 32771297-32771297 on Build GRCh38
rs200907113 Chr.1: 32771112-32771112 on Build GRCh38
rs756267925 Chr.1: 32771230-32771230 on Build GRCh38
rs752730001 Chr.1: 32771106-32771106 on Build GRCh38
rs372284962 Chr.1: 32771097-32771097 on Build GRCh38
rs115626337 Chr.1: 32771251-32771251 on Build GRCh38
rs755334396 Chr.1: 32771231-32771231 on Build GRCh38
rs781470007 Chr.1: 32771234-32771234 on Build GRCh38
rs969632302 Chr.1: 32771241-32771241 on Build GRCh38
rs770523995 Chr.1: 32771259-32771259 on Build GRCh38
rs200468241 Chr.1: 32771308-32771308 on Build GRCh38
rs775364916 Chr.1: 32771286-32771286 on Build GRCh38
rs774186643 Chr.1: 32771200-32771200 on Build GRCh38
rs748528154 Chr.1: 32771238-32771238 on Build GRCh38
rs770340335 Chr.1: 32771171-32771171 on Build GRCh38

View Full Product Details