Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30645407-30645634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2E2
Entrez Gene ID
2961
Gene Name
general transcription factor IIE subunit 2
Gene Aliases
FE, TF2E2, TFIIE-B, TTD6
UniGene
Hs.77100
Gene Chromosome Location
Chr.8: 30578514-30658241 on Build GRCh38
Transcripts
NM_002095.4 XM_011544509.2 XM_011544510.1 XM_017013363.1 XM_017013364.1 NM_001206847.1 XM_017012943.1

Gene Symbol
SMIM18
Entrez Gene ID
100507341
Gene Name
small integral membrane protein 18
Gene Aliases
-
UniGene
Hs.659493
Gene Chromosome Location
Chr.8: 30637771-30645956 on Build GRCh38
Transcripts
NM_002095.4 XM_011544509.2 XM_011544510.1 XM_017013363.1 XM_017013364.1 NM_001206847.1 XM_017012943.1

SNP Detail

SNP ID Chromosome Location
rs942345199 Chr.8: 30645517-30645517 on Build GRCh38
rs1044045147 Chr.8: 30645528-30645528 on Build GRCh38
rs1039436776 Chr.8: 30645526-30645526 on Build GRCh38
rs779604816 Chr.8: 30645532-30645532 on Build GRCh38
rs377468023 Chr.8: 30645554-30645554 on Build GRCh38
rs748620452 Chr.8: 30645448-30645448 on Build GRCh38
rs909575093 Chr.8: 30645516-30645516 on Build GRCh38
rs746606291 Chr.8: 30645577-30645577 on Build GRCh38
rs188901748 Chr.8: 30645458-30645458 on Build GRCh38
rs933733444 Chr.8: 30645597-30645597 on Build GRCh38
rs984995372 Chr.8: 30645484-30645484 on Build GRCh38
rs934491983 Chr.8: 30645471-30645471 on Build GRCh38
rs529043232 Chr.8: 30645496-30645496 on Build GRCh38
rs145837810 Chr.8: 30645519-30645519 on Build GRCh38
rs1052895547 Chr.8: 30645501-30645501 on Build GRCh38
rs539320642 Chr.8: 30645600-30645601 on Build GRCh38
rs757242613 Chr.8: 30645453-30645453 on Build GRCh38
rs745410470 Chr.8: 30645476-30645476 on Build GRCh38
rs892883845 Chr.8: 30645515-30645515 on Build GRCh38

View Full Product Details