Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49444570-49444831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
UniGene
Hs.485527
Gene Chromosome Location
Chr.6: 49430360-49463328 on Build GRCh38
Transcripts
NM_000255.3 XM_005249143.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425900 c.1561-1G>A p.? Chr.6: 49444755-49444755 on Build GRCh38
5700257 c.1566A>T p.K522N Chr.6: 49444749-49444749 on Build GRCh38
5036858 c.1579C>G p.Q527E Chr.6: 49444736-49444736 on Build GRCh38
6718274 c.1669C>T p.R557W Chr.6: 49444646-49444646 on Build GRCh38
5018226 c.1622C>T p.A541V Chr.6: 49444693-49444693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765607098 Chr.6: 49444764-49444765 on Build GRCh38
rs777199774 Chr.6: 49444605-49444605 on Build GRCh38
rs750355021 Chr.6: 49444732-49444732 on Build GRCh38
rs765713578 Chr.6: 49444622-49444622 on Build GRCh38
rs753512817 Chr.6: 49444718-49444718 on Build GRCh38
rs574196935 Chr.6: 49444768-49444768 on Build GRCh38
rs770086093 Chr.6: 49444692-49444692 on Build GRCh38
rs764985110 Chr.6: 49444721-49444721 on Build GRCh38
rs199886805 Chr.6: 49444629-49444629 on Build GRCh38
rs776320348 Chr.6: 49444786-49444786 on Build GRCh38
rs1012591559 Chr.6: 49444792-49444792 on Build GRCh38
rs570658958 Chr.6: 49444747-49444747 on Build GRCh38
rs756389945 Chr.6: 49444645-49444645 on Build GRCh38
rs566802616 Chr.6: 49444606-49444606 on Build GRCh38
rs759790687 Chr.6: 49444633-49444644 on Build GRCh38
rs372988725 Chr.6: 49444745-49444745 on Build GRCh38
rs745828854 Chr.6: 49444668-49444668 on Build GRCh38
rs144665233 Chr.6: 49444750-49444750 on Build GRCh38
rs758475503 Chr.6: 49444671-49444671 on Build GRCh38
rs764316430 Chr.6: 49444656-49444656 on Build GRCh38
rs377427994 Chr.6: 49444630-49444630 on Build GRCh38
rs760183775 Chr.6: 49444712-49444712 on Build GRCh38
rs541859735 Chr.6: 49444670-49444670 on Build GRCh38
rs886061559 Chr.6: 49444646-49444646 on Build GRCh38
rs748358807 Chr.6: 49444751-49444751 on Build GRCh38
rs748492928 Chr.6: 49444690-49444690 on Build GRCh38
rs768189807 Chr.6: 49444785-49444785 on Build GRCh38
rs1012559525 Chr.6: 49444661-49444661 on Build GRCh38
rs760130347 Chr.6: 49444783-49444783 on Build GRCh38
rs774853621 Chr.6: 49444778-49444778 on Build GRCh38
rs796052008 Chr.6: 49444684-49444685 on Build GRCh38
rs763417266 Chr.6: 49444700-49444700 on Build GRCh38
rs753564352 Chr.6: 49444652-49444652 on Build GRCh38
rs886042128 Chr.6: 49444716-49444716 on Build GRCh38
rs752322747 Chr.6: 49444636-49444636 on Build GRCh38
rs755824133 Chr.6: 49444643-49444643 on Build GRCh38
rs139765234 Chr.6: 49444717-49444717 on Build GRCh38
rs78150750 Chr.6: 49444685-49444685 on Build GRCh38
rs773738310 Chr.6: 49444693-49444693 on Build GRCh38
rs150642856 Chr.6: 49444686-49444686 on Build GRCh38
rs752706245 Chr.6: 49444766-49444766 on Build GRCh38
rs202102006 Chr.6: 49444795-49444795 on Build GRCh38
rs761423398 Chr.6: 49444789-49444789 on Build GRCh38
rs912821000 Chr.6: 49444731-49444731 on Build GRCh38
rs771363186 Chr.6: 49444774-49444774 on Build GRCh38
rs778864344 Chr.6: 49444666-49444666 on Build GRCh38
rs1141321 Chr.6: 49444720-49444720 on Build GRCh38
rs1054219157 Chr.6: 49444777-49444777 on Build GRCh38
rs760537903 Chr.6: 49444650-49444650 on Build GRCh38
rs780334099 Chr.6: 49444676-49444676 on Build GRCh38
rs879253845 Chr.6: 49444660-49444660 on Build GRCh38
rs369902876 Chr.6: 49444708-49444708 on Build GRCh38
rs777001813 Chr.6: 49444687-49444687 on Build GRCh38
rs555905807 Chr.6: 49444758-49444758 on Build GRCh38
rs766999822 Chr.6: 49444701-49444701 on Build GRCh38
rs766976599 Chr.6: 49444634-49444634 on Build GRCh38

View Full Product Details