Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77287066-77287339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK5
Entrez Gene ID
26289
Gene Name
adenylate kinase 5
Gene Aliases
AK6
UniGene
Hs.559718
Gene Chromosome Location
Chr.1: 77281908-77559969 on Build GRCh38
Transcripts
NM_012093.3 NM_174858.2 XM_005270739.4 XM_006710572.3 XM_017001008.1 XM_017001009.1 XM_017001010.1 XM_017001011.1 XM_017001012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295786 c.192G>T p.K64N Chr.1: 77287072-77287072 on Build GRCh38
1560491 c.227G>A p.R76Q Chr.1: 77287107-77287107 on Build GRCh38
3700796 c.230G>A p.R77K Chr.1: 77287110-77287110 on Build GRCh38
7091452 c.246T>G p.N82K Chr.1: 77287126-77287126 on Build GRCh38
4532395 c.187G>A p.E63K Chr.1: 77287067-77287067 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376441818 Chr.1: 77287159-77287159 on Build GRCh38
rs201693741 Chr.1: 77287112-77287112 on Build GRCh38
rs200751298 Chr.1: 77287161-77287161 on Build GRCh38
rs942119853 Chr.1: 77287307-77287307 on Build GRCh38
rs751153431 Chr.1: 77287107-77287107 on Build GRCh38
rs762683033 Chr.1: 77287093-77287093 on Build GRCh38
rs763885359 Chr.1: 77287106-77287106 on Build GRCh38
rs988651925 Chr.1: 77287202-77287202 on Build GRCh38
rs747102066 Chr.1: 77287166-77287166 on Build GRCh38
rs756612766 Chr.1: 77287126-77287126 on Build GRCh38
rs888639149 Chr.1: 77287266-77287266 on Build GRCh38
rs949697991 Chr.1: 77287279-77287279 on Build GRCh38
rs560926484 Chr.1: 77287207-77287207 on Build GRCh38
rs777891125 Chr.1: 77287165-77287165 on Build GRCh38
rs918217046 Chr.1: 77287244-77287244 on Build GRCh38
rs751139308 Chr.1: 77287280-77287280 on Build GRCh38
rs1167204 Chr.1: 77287311-77287311 on Build GRCh38
rs766951395 Chr.1: 77287137-77287137 on Build GRCh38
rs755377990 Chr.1: 77287190-77287190 on Build GRCh38
rs546673701 Chr.1: 77287226-77287226 on Build GRCh38
rs754418678 Chr.1: 77287148-77287148 on Build GRCh38
rs752232046 Chr.1: 77287151-77287152 on Build GRCh38
rs761435726 Chr.1: 77287110-77287110 on Build GRCh38
rs370596240 Chr.1: 77287168-77287168 on Build GRCh38
rs386632621 Chr.1: 77287237-77287252 on Build GRCh38
rs918145015 Chr.1: 77287253-77287253 on Build GRCh38
rs749949045 Chr.1: 77287114-77287114 on Build GRCh38
rs755339144 Chr.1: 77287158-77287158 on Build GRCh38
rs937622024 Chr.1: 77287227-77287227 on Build GRCh38
rs1021234800 Chr.1: 77287164-77287164 on Build GRCh38
rs976706299 Chr.1: 77287139-77287139 on Build GRCh38
rs4033036 Chr.1: 77287291-77287291 on Build GRCh38
rs775266265 Chr.1: 77287236-77287237 on Build GRCh38
rs755941619 Chr.1: 77287153-77287154 on Build GRCh38
rs758609125 Chr.1: 77287162-77287162 on Build GRCh38
rs113213379 Chr.1: 77287237-77287237 on Build GRCh38
rs528202509 Chr.1: 77287221-77287221 on Build GRCh38
rs886647967 Chr.1: 77287301-77287301 on Build GRCh38
rs749226134 Chr.1: 77287171-77287171 on Build GRCh38
rs775344958 Chr.1: 77287169-77287169 on Build GRCh38
rs4033037 Chr.1: 77287252-77287252 on Build GRCh38

View Full Product Details