Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110198425-110198694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMMECR1
Entrez Gene ID
9949
Gene Name
Alport syndrome, mental retardation, midface hypoplasia and elliptocytosis chromosomal region gene 1
Gene Aliases
AMMERC1
UniGene
Hs.656243
Gene Chromosome Location
Chr.X: 110194186-110440233 on Build GRCh38
Transcripts
NM_001025580.1 NM_001171689.1 NM_015365.2

SNP Detail

SNP ID Chromosome Location
rs780387914 Chr.X: 110198513-110198513 on Build GRCh38
rs766604767 Chr.X: 110198637-110198640 on Build GRCh38
rs770217553 Chr.X: 110198510-110198510 on Build GRCh38
rs1019780827 Chr.X: 110198548-110198548 on Build GRCh38
rs765118261 Chr.X: 110198565-110198565 on Build GRCh38
rs752552594 Chr.X: 110198577-110198577 on Build GRCh38
rs756839327 Chr.X: 110198603-110198603 on Build GRCh38
rs770909242 Chr.X: 110198662-110198662 on Build GRCh38
rs373583522 Chr.X: 110198497-110198497 on Build GRCh38
rs762023059 Chr.X: 110198541-110198541 on Build GRCh38
rs771788617 Chr.X: 110198542-110198542 on Build GRCh38
rs1007948031 Chr.X: 110198505-110198505 on Build GRCh38
rs774473904 Chr.X: 110198523-110198523 on Build GRCh38
rs150423719 Chr.X: 110198649-110198649 on Build GRCh38
rs780943588 Chr.X: 110198629-110198629 on Build GRCh38
rs760812340 Chr.X: 110198571-110198573 on Build GRCh38
rs145206893 Chr.X: 110198591-110198591 on Build GRCh38
rs140247782 Chr.X: 110198562-110198562 on Build GRCh38
rs372054643 Chr.X: 110198557-110198557 on Build GRCh38
rs180715560 Chr.X: 110198517-110198517 on Build GRCh38
rs763040190 Chr.X: 110198584-110198584 on Build GRCh38
rs751709821 Chr.X: 110198460-110198460 on Build GRCh38
rs751046044 Chr.X: 110198600-110198600 on Build GRCh38
rs780517440 Chr.X: 110198668-110198668 on Build GRCh38

View Full Product Details