Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51951729-51951954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
UniGene
Hs.438918
Gene Chromosome Location
Chr.12: 51951667-51997079 on Build GRCh38
Transcripts
NM_004302.4 NM_020328.3 XM_011538966.2 XM_011538967.2 XM_017020199.1 XM_017020200.1 XM_017020201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4508884 c.78C>T p.P26P Chr.12: 51951821-51951821 on Build GRCh38
6295679 c.51G>T p.L17L Chr.12: 51951794-51951794 on Build GRCh38
7315003 c.70T>G p.S24A Chr.12: 51951813-51951813 on Build GRCh38
4659195 c.80delG p.V29fs*57 Chr.12: 51951823-51951823 on Build GRCh38
7315405 c.90G>T p.Q30H Chr.12: 51951833-51951833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374624845 Chr.12: 51951809-51951810 on Build GRCh38
rs915553529 Chr.12: 51951867-51951867 on Build GRCh38
rs745908065 Chr.12: 51951758-51951758 on Build GRCh38
rs929774571 Chr.12: 51951823-51951823 on Build GRCh38
rs781239014 Chr.12: 51951813-51951813 on Build GRCh38
rs778921623 Chr.12: 51951808-51951808 on Build GRCh38
rs772982607 Chr.12: 51951851-51951851 on Build GRCh38
rs753820498 Chr.12: 51951798-51951798 on Build GRCh38
rs1003205101 Chr.12: 51951806-51951806 on Build GRCh38
rs767042035 Chr.12: 51951773-51951773 on Build GRCh38
rs979338156 Chr.12: 51951924-51951924 on Build GRCh38
rs968941282 Chr.12: 51951901-51951901 on Build GRCh38
rs561454207 Chr.12: 51951854-51951854 on Build GRCh38
rs762262503 Chr.12: 51951770-51951770 on Build GRCh38
rs377440332 Chr.12: 51951766-51951766 on Build GRCh38
rs767970155 Chr.12: 51951850-51951850 on Build GRCh38
rs753338862 Chr.12: 51951875-51951875 on Build GRCh38
rs769911875 Chr.12: 51951762-51951762 on Build GRCh38
rs867982386 Chr.12: 51951900-51951900 on Build GRCh38
rs750134234 Chr.12: 51951778-51951778 on Build GRCh38
rs909609650 Chr.12: 51951832-51951832 on Build GRCh38
rs747847374 Chr.12: 51951809-51951809 on Build GRCh38
rs754892871 Chr.12: 51951801-51951801 on Build GRCh38
rs774956562 Chr.12: 51951848-51951848 on Build GRCh38
rs956345399 Chr.12: 51951854-51951855 on Build GRCh38
rs941094068 Chr.12: 51951836-51951836 on Build GRCh38
rs982507175 Chr.12: 51951829-51951829 on Build GRCh38
rs774523980 Chr.12: 51951769-51951769 on Build GRCh38
rs775483893 Chr.12: 51951825-51951825 on Build GRCh38
rs950369465 Chr.12: 51951895-51951895 on Build GRCh38
rs781256272 Chr.12: 51951803-51951813 on Build GRCh38
rs766040681 Chr.12: 51951797-51951797 on Build GRCh38
rs763520082 Chr.12: 51951765-51951765 on Build GRCh38
rs754557218 Chr.12: 51951880-51951880 on Build GRCh38
rs762056742 Chr.12: 51951849-51951849 on Build GRCh38
rs768997035 Chr.12: 51951847-51951847 on Build GRCh38
rs746148709 Chr.12: 51951815-51951815 on Build GRCh38
rs373237304 Chr.12: 51951754-51951754 on Build GRCh38
rs758243949 Chr.12: 51951810-51951810 on Build GRCh38
rs755660131 Chr.12: 51951779-51951779 on Build GRCh38
rs769860834 Chr.12: 51951818-51951818 on Build GRCh38
rs766132478 Chr.12: 51951858-51951858 on Build GRCh38
rs1020936784 Chr.12: 51951893-51951893 on Build GRCh38
rs749385229 Chr.12: 51951828-51951828 on Build GRCh38

View Full Product Details