Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111456119-111456344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5F1
Entrez Gene ID
515
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
Gene Aliases
PIG47
UniGene
Hs.514870
Gene Chromosome Location
Chr.1: 111449121-111461918 on Build GRCh38
Transcripts
NM_001688.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172436 c.298T>C p.F100L Chr.1: 111456160-111456160 on Build GRCh38
6057539 c.332A>G p.Y111C Chr.1: 111456194-111456194 on Build GRCh38
5469516 c.288C>T p.S96S Chr.1: 111456150-111456150 on Build GRCh38
5432421 c.322G>T p.V108L Chr.1: 111456184-111456184 on Build GRCh38
5377190 c.261T>A p.A87A Chr.1: 111456123-111456123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754949769 Chr.1: 111456192-111456192 on Build GRCh38
rs769097267 Chr.1: 111456263-111456263 on Build GRCh38
rs141114115 Chr.1: 111456301-111456301 on Build GRCh38
rs780628556 Chr.1: 111456260-111456260 on Build GRCh38
rs752631080 Chr.1: 111456225-111456225 on Build GRCh38
rs370896481 Chr.1: 111456154-111456154 on Build GRCh38
rs935082748 Chr.1: 111456245-111456245 on Build GRCh38
rs376770527 Chr.1: 111456153-111456153 on Build GRCh38
rs373086502 Chr.1: 111456259-111456259 on Build GRCh38
rs750616843 Chr.1: 111456163-111456163 on Build GRCh38
rs778799728 Chr.1: 111456218-111456218 on Build GRCh38
rs1053525580 Chr.1: 111456193-111456193 on Build GRCh38
rs767597893 Chr.1: 111456156-111456156 on Build GRCh38
rs766220727 Chr.1: 111456174-111456174 on Build GRCh38
rs774754988 Chr.1: 111456272-111456272 on Build GRCh38
rs753627949 Chr.1: 111456184-111456184 on Build GRCh38
rs17849983 Chr.1: 111456213-111456213 on Build GRCh38
rs17027699 Chr.1: 111456292-111456292 on Build GRCh38
rs148039333 Chr.1: 111456150-111456150 on Build GRCh38
rs927691716 Chr.1: 111456171-111456171 on Build GRCh38
rs759699044 Chr.1: 111456194-111456194 on Build GRCh38
rs777269751 Chr.1: 111456228-111456228 on Build GRCh38
rs746652229 Chr.1: 111456250-111456250 on Build GRCh38
rs749635122 Chr.1: 111456261-111456261 on Build GRCh38
rs946210849 Chr.1: 111456264-111456264 on Build GRCh38
rs774610006 Chr.1: 111456151-111456151 on Build GRCh38
rs980372697 Chr.1: 111456165-111456165 on Build GRCh38
rs746088793 Chr.1: 111456297-111456298 on Build GRCh38
rs11553282 Chr.1: 111456166-111456166 on Build GRCh38
rs572819940 Chr.1: 111456227-111456227 on Build GRCh38
rs577689164 Chr.1: 111456305-111456305 on Build GRCh38
rs371319407 Chr.1: 111456223-111456223 on Build GRCh38

View Full Product Details