Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29054191-29054436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA2
Entrez Gene ID
321
Gene Name
amyloid beta precursor protein binding family A member 2
Gene Aliases
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
UniGene
Hs.618112
Gene Chromosome Location
Chr.15: 28885479-29118315 on Build GRCh38
Transcripts
NM_001130414.1 NM_005503.3 XM_011521488.2 XM_011521489.2 XM_011521490.2 XM_011521491.1 XM_011521492.2 XM_011521493.2 XM_011521494.2 XM_017022101.1 XM_017022102.1 XM_017022103.1 XM_017022104.1 XM_017022105.1 XM_017022106.1 XM_017022107.1 XM_017022108.1 XM_017022109.1 XM_017022110.1 XM_017022111.1 XM_017022112.1 XM_017022113.1 XM_017022114.1 XM_017022115.1 XM_017022116.1 XM_017022117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601868 c.451G>T p.G151C Chr.15: 29054335-29054335 on Build GRCh38
7157622 c.334G>C p.E112Q Chr.15: 29054218-29054218 on Build GRCh38
3690318 c.489C>T p.G163G Chr.15: 29054373-29054373 on Build GRCh38
1470741 c.321C>T p.D107D Chr.15: 29054205-29054205 on Build GRCh38
4551837 c.541G>A p.E181K Chr.15: 29054425-29054425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760419830 Chr.15: 29054329-29054329 on Build GRCh38
rs773558118 Chr.15: 29054371-29054371 on Build GRCh38
rs572236612 Chr.15: 29054395-29054395 on Build GRCh38
rs763865869 Chr.15: 29054388-29054388 on Build GRCh38
rs372649988 Chr.15: 29054370-29054370 on Build GRCh38
rs774589846 Chr.15: 29054211-29054211 on Build GRCh38
rs754615209 Chr.15: 29054268-29054268 on Build GRCh38
rs78253846 Chr.15: 29054327-29054327 on Build GRCh38
rs750416280 Chr.15: 29054335-29054335 on Build GRCh38
rs199510860 Chr.15: 29054356-29054356 on Build GRCh38
rs569922946 Chr.15: 29054377-29054377 on Build GRCh38
rs772215906 Chr.15: 29054235-29054235 on Build GRCh38
rs375569614 Chr.15: 29054325-29054325 on Build GRCh38
rs746039722 Chr.15: 29054225-29054225 on Build GRCh38
rs765759759 Chr.15: 29054318-29054318 on Build GRCh38
rs775876729 Chr.15: 29054238-29054238 on Build GRCh38
rs749183539 Chr.15: 29054284-29054284 on Build GRCh38
rs377361968 Chr.15: 29054340-29054340 on Build GRCh38
rs369913636 Chr.15: 29054295-29054295 on Build GRCh38
rs755831568 Chr.15: 29054277-29054277 on Build GRCh38
rs148660621 Chr.15: 29054316-29054316 on Build GRCh38
rs775073176 Chr.15: 29054380-29054380 on Build GRCh38
rs372287836 Chr.15: 29054319-29054319 on Build GRCh38
rs201879619 Chr.15: 29054310-29054310 on Build GRCh38
rs199963564 Chr.15: 29054309-29054309 on Build GRCh38
rs150850452 Chr.15: 29054347-29054347 on Build GRCh38
rs373964837 Chr.15: 29054332-29054332 on Build GRCh38
rs758405411 Chr.15: 29054337-29054337 on Build GRCh38
rs148760039 Chr.15: 29054387-29054387 on Build GRCh38
rs554784948 Chr.15: 29054404-29054404 on Build GRCh38
rs765816883 Chr.15: 29054261-29054261 on Build GRCh38
rs926302102 Chr.15: 29054320-29054320 on Build GRCh38
rs111568652 Chr.15: 29054241-29054241 on Build GRCh38
rs777117052 Chr.15: 29054242-29054242 on Build GRCh38
rs149482618 Chr.15: 29054334-29054334 on Build GRCh38
rs368945995 Chr.15: 29054367-29054367 on Build GRCh38
rs144358193 Chr.15: 29054376-29054376 on Build GRCh38
rs772243386 Chr.15: 29054296-29054296 on Build GRCh38
rs143649138 Chr.15: 29054294-29054294 on Build GRCh38
rs758943858 Chr.15: 29054324-29054324 on Build GRCh38
rs747140897 Chr.15: 29054341-29054341 on Build GRCh38
rs137995892 Chr.15: 29054265-29054265 on Build GRCh38
rs144066720 Chr.15: 29054315-29054315 on Build GRCh38
rs752440788 Chr.15: 29054269-29054269 on Build GRCh38
rs760926237 Chr.15: 29054239-29054239 on Build GRCh38
rs201436314 Chr.15: 29054393-29054393 on Build GRCh38
rs762364817 Chr.15: 29054258-29054258 on Build GRCh38
rs931835068 Chr.15: 29054322-29054322 on Build GRCh38
rs777772001 Chr.15: 29054282-29054282 on Build GRCh38
rs16955009 Chr.15: 29054403-29054403 on Build GRCh38
rs578050970 Chr.15: 29054328-29054328 on Build GRCh38

View Full Product Details