Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97998503-97998760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANP32B
Entrez Gene ID
10541
Gene Name
acidic nuclear phosphoprotein 32 family member B
Gene Aliases
APRIL, PHAPI2, SSP29
UniGene
Hs.730654
Gene Chromosome Location
Chr.9: 97983207-98015943 on Build GRCh38
Transcripts
NM_006401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6315157 c.289G>T p.G97* Chr.9: 97998640-97998640 on Build GRCh38
4855697 c.241G>C p.D81H Chr.9: 97998592-97998592 on Build GRCh38
1103078 c.228C>A p.I76I Chr.9: 97998579-97998579 on Build GRCh38
296548 c.285A>G p.L95L Chr.9: 97998636-97998636 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138669342 Chr.9: 97998562-97998562 on Build GRCh38
rs768604325 Chr.9: 97998728-97998728 on Build GRCh38
rs535530800 Chr.9: 97998660-97998660 on Build GRCh38
rs991523817 Chr.9: 97998586-97998586 on Build GRCh38
rs770733850 Chr.9: 97998707-97998707 on Build GRCh38
rs759567159 Chr.9: 97998724-97998724 on Build GRCh38
rs766236757 Chr.9: 97998540-97998540 on Build GRCh38
rs774134162 Chr.9: 97998709-97998709 on Build GRCh38
rs775271168 Chr.9: 97998626-97998627 on Build GRCh38
rs552493990 Chr.9: 97998582-97998582 on Build GRCh38
rs758841211 Chr.9: 97998622-97998622 on Build GRCh38
rs914567050 Chr.9: 97998597-97998597 on Build GRCh38
rs369938387 Chr.9: 97998688-97998688 on Build GRCh38
rs756237780 Chr.9: 97998716-97998721 on Build GRCh38
rs760622494 Chr.9: 97998548-97998548 on Build GRCh38
rs769575026 Chr.9: 97998624-97998625 on Build GRCh38
rs747577407 Chr.9: 97998634-97998634 on Build GRCh38
rs754411628 Chr.9: 97998654-97998654 on Build GRCh38
rs769376133 Chr.9: 97998689-97998689 on Build GRCh38
rs749041246 Chr.9: 97998704-97998704 on Build GRCh38
rs774376504 Chr.9: 97998545-97998545 on Build GRCh38
rs753976201 Chr.9: 97998601-97998601 on Build GRCh38
rs761968017 Chr.9: 97998687-97998688 on Build GRCh38
rs757567396 Chr.9: 97998605-97998605 on Build GRCh38
rs777405753 Chr.9: 97998694-97998694 on Build GRCh38
rs750761331 Chr.9: 97998614-97998614 on Build GRCh38
rs765686344 Chr.9: 97998611-97998611 on Build GRCh38
rs61757702 Chr.9: 97998630-97998630 on Build GRCh38
rs967950783 Chr.9: 97998608-97998608 on Build GRCh38

View Full Product Details