Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184187894-184188149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF3
Entrez Gene ID
55324
Gene Name
ATP binding cassette subfamily F member 3
Gene Aliases
EST201864
UniGene
Hs.361323
Gene Chromosome Location
Chr.3: 184186075-184194012 on Build GRCh38
Transcripts
NM_018358.2 XM_011512959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7191550 c.527A>G p.Y176C Chr.3: 184187941-184187941 on Build GRCh38
4499622 c.546C>T p.N182N Chr.3: 184187960-184187960 on Build GRCh38
4115674 c.481A>G p.S161G Chr.3: 184187895-184187895 on Build GRCh38
7142350 c.536G>T p.R179L Chr.3: 184187950-184187950 on Build GRCh38
446078 c.541G>C p.E181Q Chr.3: 184187955-184187955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766606565 Chr.3: 184187938-184187938 on Build GRCh38
rs1037016398 Chr.3: 184187945-184187945 on Build GRCh38
rs201288644 Chr.3: 184187917-184187917 on Build GRCh38
rs746854914 Chr.3: 184188012-184188012 on Build GRCh38
rs780021071 Chr.3: 184188002-184188002 on Build GRCh38
rs770918133 Chr.3: 184188092-184188092 on Build GRCh38
rs147647724 Chr.3: 184187922-184187922 on Build GRCh38
rs763066511 Chr.3: 184188032-184188032 on Build GRCh38
rs753152768 Chr.3: 184187971-184187971 on Build GRCh38
rs371800074 Chr.3: 184188017-184188017 on Build GRCh38
rs953899244 Chr.3: 184188075-184188075 on Build GRCh38
rs374459635 Chr.3: 184187998-184187998 on Build GRCh38
rs751740928 Chr.3: 184187963-184187963 on Build GRCh38
rs981749323 Chr.3: 184188117-184188117 on Build GRCh38
rs768706872 Chr.3: 184188025-184188025 on Build GRCh38
rs970656020 Chr.3: 184188111-184188111 on Build GRCh38
rs563690104 Chr.3: 184188066-184188066 on Build GRCh38
rs763189066 Chr.3: 184187923-184187923 on Build GRCh38
rs778292936 Chr.3: 184187980-184187980 on Build GRCh38
rs776763989 Chr.3: 184188030-184188030 on Build GRCh38
rs767828384 Chr.3: 184188098-184188098 on Build GRCh38
rs1028690835 Chr.3: 184188056-184188056 on Build GRCh38
rs550666332 Chr.3: 184187960-184187960 on Build GRCh38
rs756776401 Chr.3: 184187989-184187989 on Build GRCh38
rs745514226 Chr.3: 184187992-184187992 on Build GRCh38
rs893627985 Chr.3: 184188063-184188063 on Build GRCh38
rs1009436981 Chr.3: 184188070-184188070 on Build GRCh38
rs774590829 Chr.3: 184188095-184188095 on Build GRCh38
rs370600302 Chr.3: 184188097-184188097 on Build GRCh38
rs756591208 Chr.3: 184187979-184187979 on Build GRCh38
rs531160654 Chr.3: 184187981-184187981 on Build GRCh38
rs142135524 Chr.3: 184187954-184187954 on Build GRCh38
rs761095198 Chr.3: 184188120-184188120 on Build GRCh38
rs767802517 Chr.3: 184187968-184187968 on Build GRCh38
rs1007607938 Chr.3: 184188028-184188030 on Build GRCh38
rs74440002 Chr.3: 184187991-184187991 on Build GRCh38
rs769949338 Chr.3: 184187918-184187918 on Build GRCh38
rs774128811 Chr.3: 184188112-184188112 on Build GRCh38
rs753029864 Chr.3: 184188100-184188100 on Build GRCh38

View Full Product Details