Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70738837-70739066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS14
Entrez Gene ID
140766
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 14
Gene Aliases
-
UniGene
Hs.352156
Gene Chromosome Location
Chr.10: 70672518-70762441 on Build GRCh38
Transcripts
NM_080722.4 NM_139155.3 XM_011539300.2 XM_011539301.2 XM_011539302.2 XM_011539303.2 XM_011539305.2 XM_011539306.2 XM_011539307.2 XM_011539308.2 XM_011539309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7084590 c.1693G>C p.G565R Chr.10: 70738926-70738926 on Build GRCh38
5969882 c.1640C>T p.S547L Chr.10: 70738873-70738873 on Build GRCh38
1675322 c.1733G>A p.R578H Chr.10: 70738966-70738966 on Build GRCh38
5969884 c.1679C>T p.S560F Chr.10: 70738912-70738912 on Build GRCh38
6530874 c.1712G>C p.C571S Chr.10: 70738945-70738945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748259077 Chr.10: 70739019-70739019 on Build GRCh38
rs774608953 Chr.10: 70739011-70739011 on Build GRCh38
rs764109367 Chr.10: 70738991-70738991 on Build GRCh38
rs267602564 Chr.10: 70738964-70738964 on Build GRCh38
rs777350272 Chr.10: 70738990-70738990 on Build GRCh38
rs528519283 Chr.10: 70738939-70738939 on Build GRCh38
rs1038391790 Chr.10: 70739022-70739022 on Build GRCh38
rs199704809 Chr.10: 70738937-70738937 on Build GRCh38
rs369100038 Chr.10: 70739020-70739020 on Build GRCh38
rs765452002 Chr.10: 70738966-70738966 on Build GRCh38
rs1024911442 Chr.10: 70738951-70738951 on Build GRCh38
rs778968867 Chr.10: 70739003-70739003 on Build GRCh38
rs752110366 Chr.10: 70738971-70738971 on Build GRCh38
rs373254498 Chr.10: 70739034-70739034 on Build GRCh38
rs1047714696 Chr.10: 70739016-70739016 on Build GRCh38
rs564750212 Chr.10: 70738952-70738952 on Build GRCh38
rs774804568 Chr.10: 70739042-70739042 on Build GRCh38
rs973580014 Chr.10: 70738909-70738909 on Build GRCh38
rs376581074 Chr.10: 70738993-70738993 on Build GRCh38
rs575503783 Chr.10: 70738881-70738881 on Build GRCh38
rs758947452 Chr.10: 70738922-70738922 on Build GRCh38
rs375988679 Chr.10: 70739041-70739041 on Build GRCh38
rs763355483 Chr.10: 70739040-70739040 on Build GRCh38
rs765747013 Chr.10: 70738924-70738924 on Build GRCh38
rs988164690 Chr.10: 70738940-70738940 on Build GRCh38
rs758624356 Chr.10: 70738972-70738972 on Build GRCh38
rs138612430 Chr.10: 70738876-70738876 on Build GRCh38
rs755829295 Chr.10: 70738944-70738944 on Build GRCh38
rs772978804 Chr.10: 70738873-70738873 on Build GRCh38
rs1005551716 Chr.10: 70738892-70738892 on Build GRCh38
rs777030272 Chr.10: 70738887-70738887 on Build GRCh38
rs1014163594 Chr.10: 70739004-70739004 on Build GRCh38
rs563546901 Chr.10: 70738880-70738880 on Build GRCh38
rs199765284 Chr.10: 70738938-70738938 on Build GRCh38
rs769679153 Chr.10: 70738857-70738857 on Build GRCh38
rs762125445 Chr.10: 70738965-70738965 on Build GRCh38
rs142896753 Chr.10: 70738967-70738967 on Build GRCh38
rs368640269 Chr.10: 70738949-70738949 on Build GRCh38
rs887694112 Chr.10: 70739018-70739018 on Build GRCh38
rs368809040 Chr.10: 70738877-70738877 on Build GRCh38
rs765656395 Chr.10: 70738891-70738891 on Build GRCh38
rs371476393 Chr.10: 70738958-70738958 on Build GRCh38
rs755808207 Chr.10: 70738982-70738982 on Build GRCh38
rs777630903 Chr.10: 70738947-70738947 on Build GRCh38
rs182084110 Chr.10: 70738890-70738890 on Build GRCh38
rs945739784 Chr.10: 70738953-70738953 on Build GRCh38
rs750854699 Chr.10: 70738914-70738914 on Build GRCh38
rs1041435605 Chr.10: 70738970-70738970 on Build GRCh38
rs749282565 Chr.10: 70738874-70738874 on Build GRCh38
rs768901348 Chr.10: 70738960-70738960 on Build GRCh38
rs766880126 Chr.10: 70738969-70738969 on Build GRCh38
rs745818058 Chr.10: 70739005-70739005 on Build GRCh38
rs776944253 Chr.10: 70738961-70738961 on Build GRCh38
rs768817178 Chr.10: 70739012-70739012 on Build GRCh38
rs752283953 Chr.10: 70738980-70738980 on Build GRCh38

View Full Product Details