Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151122656-151122881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
UniGene
Hs.647075
Gene Chromosome Location
Chr.7: 151085831-151144436 on Build GRCh38
Transcripts
NM_001042535.3 NM_001281300.1 NM_001308304.1 NM_001308305.1 NM_031946.6 XM_005249943.2 XM_011515780.1 XM_017011732.1 XM_017011733.1 XM_017011734.1 XM_017011735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280335 c.1135G>A p.A379T Chr.7: 151122731-151122731 on Build GRCh38
5526403 c.1179C>T p.L393L Chr.7: 151122775-151122775 on Build GRCh38
452639 c.1183C>T p.P395S Chr.7: 151122779-151122779 on Build GRCh38
6596072 c.1146C>A p.S382S Chr.7: 151122742-151122742 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771393307 Chr.7: 151122702-151122702 on Build GRCh38
rs746246221 Chr.7: 151122709-151122709 on Build GRCh38
rs377520215 Chr.7: 151122687-151122687 on Build GRCh38
rs765340754 Chr.7: 151122834-151122834 on Build GRCh38
rs768258267 Chr.7: 151122769-151122769 on Build GRCh38
rs371190738 Chr.7: 151122704-151122704 on Build GRCh38
rs761802545 Chr.7: 151122778-151122778 on Build GRCh38
rs373004175 Chr.7: 151122775-151122775 on Build GRCh38
rs747568465 Chr.7: 151122695-151122695 on Build GRCh38
rs1043237949 Chr.7: 151122765-151122765 on Build GRCh38
rs762318311 Chr.7: 151122749-151122749 on Build GRCh38
rs904264993 Chr.7: 151122805-151122805 on Build GRCh38
rs1001659628 Chr.7: 151122856-151122856 on Build GRCh38
rs767290027 Chr.7: 151122806-151122806 on Build GRCh38
rs761625835 Chr.7: 151122835-151122845 on Build GRCh38
rs769262330 Chr.7: 151122714-151122714 on Build GRCh38
rs752657020 Chr.7: 151122675-151122675 on Build GRCh38
rs904523024 Chr.7: 151122796-151122796 on Build GRCh38
rs758540596 Chr.7: 151122676-151122676 on Build GRCh38
rs946100216 Chr.7: 151122698-151122698 on Build GRCh38
rs775238838 Chr.7: 151122741-151122741 on Build GRCh38
rs760527618 Chr.7: 151122828-151122828 on Build GRCh38
rs750163827 Chr.7: 151122817-151122817 on Build GRCh38
rs778093330 Chr.7: 151122685-151122685 on Build GRCh38
rs1043198163 Chr.7: 151122748-151122748 on Build GRCh38
rs866481933 Chr.7: 151122764-151122764 on Build GRCh38

View Full Product Details