Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49950742-49951001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP2
Entrez Gene ID
359
Gene Name
aquaporin 2
Gene Aliases
AQP-CD, WCH-CD
UniGene
Hs.130730
Gene Chromosome Location
Chr.12: 49950741-49958881 on Build GRCh38
Transcripts
NM_000486.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6602572 c.171G>T p.Q57H Chr.12: 49951001-49951001 on Build GRCh38
6602571 c.141G>A p.A47A Chr.12: 49950971-49950971 on Build GRCh38
5051664 c.114G>A p.L38L Chr.12: 49950944-49950944 on Build GRCh38
3461769 c.13C>T p.R5C Chr.12: 49950843-49950843 on Build GRCh38
1245258 c.73T>G p.F25V Chr.12: 49950903-49950903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964170650 Chr.12: 49950915-49950915 on Build GRCh38
rs779085450 Chr.12: 49950899-49950899 on Build GRCh38
rs563444539 Chr.12: 49950771-49950771 on Build GRCh38
rs775148085 Chr.12: 49950872-49950872 on Build GRCh38
rs879038380 Chr.12: 49950766-49950766 on Build GRCh38
rs769345320 Chr.12: 49950941-49950941 on Build GRCh38
rs62620006 Chr.12: 49950772-49950772 on Build GRCh38
rs373926562 Chr.12: 49950914-49950914 on Build GRCh38
rs143249649 Chr.12: 49950765-49950765 on Build GRCh38
rs970041953 Chr.12: 49950779-49950779 on Build GRCh38
rs778267384 Chr.12: 49950909-49950909 on Build GRCh38
rs140262864 Chr.12: 49950971-49950971 on Build GRCh38
rs187754589 Chr.12: 49950857-49950857 on Build GRCh38
rs771261931 Chr.12: 49950923-49950923 on Build GRCh38
rs762574510 Chr.12: 49950873-49950873 on Build GRCh38
rs747971391 Chr.12: 49950891-49950893 on Build GRCh38
rs139709797 Chr.12: 49950917-49950917 on Build GRCh38
rs372883230 Chr.12: 49950841-49950841 on Build GRCh38
rs762664471 Chr.12: 49950955-49950955 on Build GRCh38
rs758221908 Chr.12: 49950794-49950794 on Build GRCh38
rs756096963 Chr.12: 49950980-49950980 on Build GRCh38
rs774952238 Chr.12: 49950944-49950944 on Build GRCh38
rs758661503 Chr.12: 49950908-49950908 on Build GRCh38
rs61733029 Chr.12: 49950869-49950869 on Build GRCh38
rs104894336 Chr.12: 49950894-49950894 on Build GRCh38
rs763727714 Chr.12: 49950958-49950958 on Build GRCh38
rs780251337 Chr.12: 49950816-49950816 on Build GRCh38
rs377296676 Chr.12: 49950881-49950881 on Build GRCh38
rs148085137 Chr.12: 49950862-49950862 on Build GRCh38
rs777139117 Chr.12: 49950860-49950860 on Build GRCh38
rs374778728 Chr.12: 49950780-49950780 on Build GRCh38
rs772201159 Chr.12: 49950919-49950941 on Build GRCh38
rs148006652 Chr.12: 49950844-49950844 on Build GRCh38
rs777498744 Chr.12: 49950805-49950805 on Build GRCh38
rs772585705 Chr.12: 49950843-49950843 on Build GRCh38
rs779279677 Chr.12: 49950837-49950837 on Build GRCh38
rs777037742 Chr.12: 49950932-49950932 on Build GRCh38
rs757280714 Chr.12: 49950891-49950891 on Build GRCh38
rs147136099 Chr.12: 49950893-49950893 on Build GRCh38
rs141361202 Chr.12: 49950783-49950783 on Build GRCh38
rs995684800 Chr.12: 49950970-49950970 on Build GRCh38
rs752281925 Chr.12: 49950784-49950784 on Build GRCh38
rs761515066 Chr.12: 49950966-49950966 on Build GRCh38
rs370614216 Chr.12: 49950821-49950821 on Build GRCh38
rs746338782 Chr.12: 49950933-49950933 on Build GRCh38
rs751311089 Chr.12: 49950885-49950885 on Build GRCh38
rs767301067 Chr.12: 49950968-49950968 on Build GRCh38
rs912994825 Chr.12: 49950822-49950822 on Build GRCh38
rs1045287969 Chr.12: 49950793-49950793 on Build GRCh38
rs761283952 Chr.12: 49950845-49950845 on Build GRCh38
rs768720147 Chr.12: 49950829-49950829 on Build GRCh38
rs200777769 Chr.12: 49950815-49950815 on Build GRCh38
rs200706192 Chr.12: 49950900-49950900 on Build GRCh38
rs748337255 Chr.12: 49950839-49950839 on Build GRCh38
rs763919973 Chr.12: 49950879-49950879 on Build GRCh38

View Full Product Details