Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154584771-154585044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAR
Entrez Gene ID
103
Gene Name
adenosine deaminase, RNA specific
Gene Aliases
ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
UniGene
Hs.12341
Gene Chromosome Location
Chr.1: 154582057-154628016 on Build GRCh38
Transcripts
NM_001025107.2 NM_001111.4 NM_001193495.1 NM_015840.3 NM_015841.3 XM_006711109.2 XM_006711111.3 XM_006711112.2 XM_006711113.2 XM_011509060.1 XM_011509061.2 XM_011509062.1 XM_017000037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6594625 c.3786G>T p.K1262N Chr.1: 154584830-154584830 on Build GRCh38
4423705 c.3712G>A p.A1238T Chr.1: 154584904-154584904 on Build GRCh38
2209206 c.3568C>T p.R1190C Chr.1: 154584919-154584919 on Build GRCh38
4022834 c.3448C>T p.R1150W Chr.1: 154585039-154585039 on Build GRCh38
6567142 c.3616C>T p.L1206F Chr.1: 154585000-154585000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768173047 Chr.1: 154584911-154584911 on Build GRCh38
rs146861068 Chr.1: 154584805-154584805 on Build GRCh38
rs867756770 Chr.1: 154584840-154584840 on Build GRCh38
rs1041447405 Chr.1: 154584992-154584992 on Build GRCh38
rs751814000 Chr.1: 154584796-154584796 on Build GRCh38
rs755264415 Chr.1: 154584968-154584968 on Build GRCh38
rs577255099 Chr.1: 154584972-154584972 on Build GRCh38
rs766835478 Chr.1: 154584962-154584962 on Build GRCh38
rs758253421 Chr.1: 154585025-154585025 on Build GRCh38
rs866441514 Chr.1: 154584966-154584966 on Build GRCh38
rs776752850 Chr.1: 154584905-154584905 on Build GRCh38
rs1030546987 Chr.1: 154584812-154584812 on Build GRCh38
rs923616616 Chr.1: 154584942-154584942 on Build GRCh38
rs761344509 Chr.1: 154584925-154584925 on Build GRCh38
rs28936681 Chr.1: 154584993-154584993 on Build GRCh38
rs146625055 Chr.1: 154584964-154584964 on Build GRCh38
rs748204293 Chr.1: 154584803-154584803 on Build GRCh38
rs1044845711 Chr.1: 154585024-154585024 on Build GRCh38
rs781379281 Chr.1: 154584800-154584800 on Build GRCh38
rs373736154 Chr.1: 154584833-154584833 on Build GRCh38
rs370464259 Chr.1: 154584920-154584920 on Build GRCh38
rs764682423 Chr.1: 154584939-154584939 on Build GRCh38
rs367899281 Chr.1: 154584906-154584906 on Build GRCh38
rs745742147 Chr.1: 154584813-154584813 on Build GRCh38
rs370314259 Chr.1: 154584799-154584799 on Build GRCh38
rs777931839 Chr.1: 154585006-154585006 on Build GRCh38
rs752914135 Chr.1: 154584973-154584973 on Build GRCh38
rs745866663 Chr.1: 154585020-154585020 on Build GRCh38
rs751778442 Chr.1: 154584814-154584814 on Build GRCh38
rs886045338 Chr.1: 154584842-154584842 on Build GRCh38
rs566390171 Chr.1: 154584983-154584983 on Build GRCh38
rs979532140 Chr.1: 154584919-154584919 on Build GRCh38
rs374282086 Chr.1: 154584946-154584946 on Build GRCh38
rs986264460 Chr.1: 154584903-154584903 on Build GRCh38
rs769951204 Chr.1: 154584804-154584804 on Build GRCh38
rs533561093 Chr.1: 154584956-154584956 on Build GRCh38
rs931510849 Chr.1: 154584824-154584824 on Build GRCh38
rs368425519 Chr.1: 154584947-154584947 on Build GRCh38

View Full Product Details