Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42745681-42745922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5004198 c.3658G>C p.G1220R Chr.15: 42745872-42745872 on Build GRCh38
3999528 c.3711G>C p.K1237N Chr.15: 42745819-42745819 on Build GRCh38
4955337 c.3693C>A p.T1231T Chr.15: 42745837-42745837 on Build GRCh38
4768862 c.3691A>C p.T1231P Chr.15: 42745839-42745839 on Build GRCh38
4449890 c.3697delC p.Q1233fs*>12 Chr.15: 42745833-42745833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057415670 Chr.15: 42745791-42745791 on Build GRCh38
rs756417911 Chr.15: 42745744-42745744 on Build GRCh38
rs1047401617 Chr.15: 42745770-42745770 on Build GRCh38
rs756285172 Chr.15: 42745816-42745817 on Build GRCh38
rs746735760 Chr.15: 42745863-42745863 on Build GRCh38
rs754081258 Chr.15: 42745825-42745826 on Build GRCh38
rs897816474 Chr.15: 42745838-42745838 on Build GRCh38
rs746256663 Chr.15: 42745832-42745833 on Build GRCh38
rs766632304 Chr.15: 42745823-42745823 on Build GRCh38
rs36104367 Chr.15: 42745808-42745808 on Build GRCh38
rs948557799 Chr.15: 42745725-42745725 on Build GRCh38
rs1044558705 Chr.15: 42745826-42745826 on Build GRCh38
rs768597439 Chr.15: 42745867-42745867 on Build GRCh38
rs752846400 Chr.15: 42745839-42745839 on Build GRCh38
rs892918562 Chr.15: 42745718-42745718 on Build GRCh38
rs771394738 Chr.15: 42745890-42745890 on Build GRCh38
rs762502316 Chr.15: 42745882-42745882 on Build GRCh38
rs778049202 Chr.15: 42745819-42745832 on Build GRCh38
rs775543788 Chr.15: 42745792-42745792 on Build GRCh38
rs200391495 Chr.15: 42745819-42745819 on Build GRCh38
rs768169944 Chr.15: 42745903-42745903 on Build GRCh38
rs372671783 Chr.15: 42745743-42745743 on Build GRCh38
rs768071683 Chr.15: 42745835-42745835 on Build GRCh38
rs772703443 Chr.15: 42745822-42745822 on Build GRCh38
rs769265704 Chr.15: 42745873-42745873 on Build GRCh38
rs776506980 Chr.15: 42745872-42745872 on Build GRCh38
rs759930908 Chr.15: 42745899-42745899 on Build GRCh38
rs755536772 Chr.15: 42745830-42745830 on Build GRCh38
rs1054598286 Chr.15: 42745708-42745709 on Build GRCh38
rs530117994 Chr.15: 42745843-42745843 on Build GRCh38
rs752168838 Chr.15: 42745828-42745828 on Build GRCh38
rs772967476 Chr.15: 42745878-42745878 on Build GRCh38
rs749447430 Chr.15: 42745850-42745850 on Build GRCh38
rs779705342 Chr.15: 42745862-42745862 on Build GRCh38
rs778095416 Chr.15: 42745849-42745849 on Build GRCh38
rs907495112 Chr.15: 42745782-42745782 on Build GRCh38
rs764895699 Chr.15: 42745820-42745820 on Build GRCh38
rs369876689 Chr.15: 42745834-42745834 on Build GRCh38
rs952126930 Chr.15: 42745871-42745871 on Build GRCh38
rs1013116237 Chr.15: 42745816-42745816 on Build GRCh38
rs149561490 Chr.15: 42745762-42745762 on Build GRCh38
rs904618809 Chr.15: 42745833-42745833 on Build GRCh38
rs768218388 Chr.15: 42745800-42745800 on Build GRCh38
rs939028506 Chr.15: 42745785-42745785 on Build GRCh38
rs779131082 Chr.15: 42745830-42745831 on Build GRCh38
rs757619489 Chr.15: 42745827-42745828 on Build GRCh38
rs74398902 Chr.15: 42745861-42745861 on Build GRCh38
rs745704884 Chr.15: 42745771-42745771 on Build GRCh38
rs771731759 Chr.15: 42745786-42745786 on Build GRCh38
rs774584978 Chr.15: 42745894-42745894 on Build GRCh38

View Full Product Details