Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114675624-114675865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD1
Entrez Gene ID
270
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
MAD, MADA, MMDD
UniGene
Hs.89570
Gene Chromosome Location
Chr.1: 114673098-114695618 on Build GRCh38
Transcripts
NM_000036.2 NM_001172626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019527 c.1673C>G p.S558C Chr.1: 114675635-114675635 on Build GRCh38
73651 c.1517T>C p.I506T Chr.1: 114675692-114675692 on Build GRCh38
4947263 c.1616T>C p.I539T Chr.1: 114675692-114675692 on Build GRCh38
6327365 c.1573T>C p.S525P Chr.1: 114675636-114675636 on Build GRCh38
3471636 c.1679G>A p.S560N Chr.1: 114675629-114675629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375744047 Chr.1: 114675732-114675732 on Build GRCh38
rs1037126459 Chr.1: 114675645-114675645 on Build GRCh38
rs534836200 Chr.1: 114675794-114675794 on Build GRCh38
rs1022651551 Chr.1: 114675808-114675808 on Build GRCh38
rs965629361 Chr.1: 114675786-114675786 on Build GRCh38
rs761014026 Chr.1: 114675654-114675654 on Build GRCh38
rs755036028 Chr.1: 114675668-114675668 on Build GRCh38
rs745329451 Chr.1: 114675711-114675711 on Build GRCh38
rs1018473855 Chr.1: 114675782-114675782 on Build GRCh38
rs765199972 Chr.1: 114675672-114675672 on Build GRCh38
rs766575532 Chr.1: 114675744-114675744 on Build GRCh38
rs760979547 Chr.1: 114675743-114675743 on Build GRCh38
rs540449477 Chr.1: 114675768-114675768 on Build GRCh38
rs752670934 Chr.1: 114675841-114675841 on Build GRCh38
rs746816406 Chr.1: 114675692-114675692 on Build GRCh38
rs766515907 Chr.1: 114675662-114675662 on Build GRCh38
rs769233668 Chr.1: 114675718-114675718 on Build GRCh38
rs756863305 Chr.1: 114675696-114675696 on Build GRCh38
rs777771945 Chr.1: 114675690-114675690 on Build GRCh38
rs555960232 Chr.1: 114675776-114675776 on Build GRCh38
rs769775942 Chr.1: 114675719-114675719 on Build GRCh38
rs759632948 Chr.1: 114675830-114675830 on Build GRCh38
rs2070986 Chr.1: 114675792-114675792 on Build GRCh38
rs776767742 Chr.1: 114675828-114675828 on Build GRCh38
rs745970850 Chr.1: 114675829-114675829 on Build GRCh38
rs771163678 Chr.1: 114675832-114675832 on Build GRCh38
rs573448158 Chr.1: 114675673-114675673 on Build GRCh38
rs558299212 Chr.1: 114675665-114675665 on Build GRCh38
rs918607025 Chr.1: 114675797-114675797 on Build GRCh38
rs773416119 Chr.1: 114675649-114675649 on Build GRCh38
rs780737293 Chr.1: 114675707-114675707 on Build GRCh38
rs371376353 Chr.1: 114675832-114675832 on Build GRCh38
rs377381561 Chr.1: 114675729-114675729 on Build GRCh38
rs768630513 Chr.1: 114675788-114675788 on Build GRCh38
rs748718508 Chr.1: 114675730-114675730 on Build GRCh38
rs773499936 Chr.1: 114675740-114675740 on Build GRCh38
rs1003243163 Chr.1: 114675671-114675671 on Build GRCh38

View Full Product Details