Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49794253-49794478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA2
Entrez Gene ID
10449
Gene Name
acetyl-CoA acyltransferase 2
Gene Aliases
DSAEC
UniGene
Hs.200136
Gene Chromosome Location
Chr.18: 49783504-49813881 on Build GRCh38
Transcripts
NM_006111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4072425 c.510A>G p.V170V Chr.18: 49794347-49794347 on Build GRCh38
7318190 c.527G>A p.R176K Chr.18: 49794330-49794330 on Build GRCh38
3526158 c.557C>T p.S186L Chr.18: 49794300-49794300 on Build GRCh38
1750533 c.564G>C p.Q188H Chr.18: 49794293-49794293 on Build GRCh38
3717922 c.520A>G p.I174V Chr.18: 49794337-49794337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761533071 Chr.18: 49794444-49794444 on Build GRCh38
rs148412743 Chr.18: 49794341-49794341 on Build GRCh38
rs771734017 Chr.18: 49794433-49794434 on Build GRCh38
rs772163589 Chr.18: 49794374-49794374 on Build GRCh38
rs762428287 Chr.18: 49794300-49794300 on Build GRCh38
rs371834662 Chr.18: 49794412-49794412 on Build GRCh38
rs748291888 Chr.18: 49794370-49794370 on Build GRCh38
rs1057076742 Chr.18: 49794376-49794376 on Build GRCh38
rs979794620 Chr.18: 49794378-49794378 on Build GRCh38
rs201606320 Chr.18: 49794391-49794391 on Build GRCh38
rs371883468 Chr.18: 49794392-49794392 on Build GRCh38
rs747748736 Chr.18: 49794340-49794341 on Build GRCh38
rs113627528 Chr.18: 49794430-49794430 on Build GRCh38
rs775944059 Chr.18: 49794414-49794414 on Build GRCh38
rs376134556 Chr.18: 49794409-49794409 on Build GRCh38
rs750176890 Chr.18: 49794326-49794326 on Build GRCh38
rs751218627 Chr.18: 49794314-49794314 on Build GRCh38
rs778423944 Chr.18: 49794331-49794331 on Build GRCh38
rs781056730 Chr.18: 49794325-49794325 on Build GRCh38
rs34783635 Chr.18: 49794342-49794342 on Build GRCh38
rs763645430 Chr.18: 49794427-49794427 on Build GRCh38
rs375055558 Chr.18: 49794425-49794425 on Build GRCh38
rs747244408 Chr.18: 49794407-49794407 on Build GRCh38
rs144829415 Chr.18: 49794315-49794315 on Build GRCh38
rs780091058 Chr.18: 49794364-49794364 on Build GRCh38
rs763693994 Chr.18: 49794310-49794310 on Build GRCh38

View Full Product Details