Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50934515-50934776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
UniGene
Hs.413366
Gene Chromosome Location
Chr.15: 50908569-51005900 on Build GRCh38
Transcripts
NM_001252127.1 NM_007347.4 XM_005254264.3 XM_006720447.3 XM_017022042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936112 c.928C>T p.H310Y Chr.15: 50934682-50934682 on Build GRCh38
4185617 c.915A>T p.R305S Chr.15: 50934669-50934669 on Build GRCh38
4185616 c.914G>T p.R305I Chr.15: 50934668-50934668 on Build GRCh38
962645 c.911G>A p.R304Q Chr.15: 50934665-50934665 on Build GRCh38
962644 c.877G>T p.E293* Chr.15: 50934631-50934631 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770155362 Chr.15: 50934556-50934558 on Build GRCh38
rs764916204 Chr.15: 50934730-50934730 on Build GRCh38
rs753372922 Chr.15: 50934705-50934705 on Build GRCh38
rs748280262 Chr.15: 50934586-50934586 on Build GRCh38
rs769721146 Chr.15: 50934589-50934589 on Build GRCh38
rs771128366 Chr.15: 50934643-50934643 on Build GRCh38
rs184106196 Chr.15: 50934584-50934584 on Build GRCh38
rs569567659 Chr.15: 50934664-50934664 on Build GRCh38
rs111666207 Chr.15: 50934651-50934651 on Build GRCh38
rs866619499 Chr.15: 50934684-50934684 on Build GRCh38
rs778905594 Chr.15: 50934567-50934567 on Build GRCh38
rs757033210 Chr.15: 50934708-50934708 on Build GRCh38
rs867200073 Chr.15: 50934572-50934572 on Build GRCh38
rs763558290 Chr.15: 50934703-50934703 on Build GRCh38
rs986910080 Chr.15: 50934646-50934646 on Build GRCh38
rs146832617 Chr.15: 50934626-50934626 on Build GRCh38
rs745650277 Chr.15: 50934583-50934583 on Build GRCh38
rs753098581 Chr.15: 50934682-50934682 on Build GRCh38
rs768086723 Chr.15: 50934665-50934665 on Build GRCh38
rs200153804 Chr.15: 50934653-50934653 on Build GRCh38
rs536656846 Chr.15: 50934686-50934686 on Build GRCh38
rs937760553 Chr.15: 50934655-50934655 on Build GRCh38
rs112147034 Chr.15: 50934630-50934630 on Build GRCh38
rs758216342 Chr.15: 50934745-50934745 on Build GRCh38
rs756945413 Chr.15: 50934551-50934551 on Build GRCh38
rs763213142 Chr.15: 50934629-50934629 on Build GRCh38

View Full Product Details