Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31616719-31616990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIF1
Entrez Gene ID
199
Gene Name
allograft inflammatory factor 1
Gene Aliases
AIF-1, IBA1, IRT-1, IRT1
UniGene
Hs.76364
Gene Chromosome Location
Chr.6: 31615209-31617025 on Build GRCh38
Transcripts
NM_001318970.1 NM_001623.4 NM_032955.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3861714 c.482C>T p.P161L Chr.6: 31616896-31616896 on Build GRCh38
342536 c.394G>A p.E132K Chr.6: 31616850-31616850 on Build GRCh38
3861712 c.448G>T p.G150C Chr.6: 31616862-31616862 on Build GRCh38
3624604 c.363C>T p.I121I Chr.6: 31616819-31616819 on Build GRCh38
7146176 c.408G>C p.L136L Chr.6: 31616822-31616822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758014733 Chr.6: 31616842-31616842 on Build GRCh38
rs368206020 Chr.6: 31616788-31616788 on Build GRCh38
rs376483694 Chr.6: 31616928-31616928 on Build GRCh38
rs898145444 Chr.6: 31616766-31616766 on Build GRCh38
rs897398615 Chr.6: 31616951-31616951 on Build GRCh38
rs763238488 Chr.6: 31616915-31616915 on Build GRCh38
rs371722539 Chr.6: 31616774-31616774 on Build GRCh38
rs746520732 Chr.6: 31616862-31616862 on Build GRCh38
rs889757709 Chr.6: 31616894-31616894 on Build GRCh38
rs1049820835 Chr.6: 31616869-31616869 on Build GRCh38
rs770235283 Chr.6: 31616907-31616907 on Build GRCh38
rs934040063 Chr.6: 31616864-31616864 on Build GRCh38
rs113340037 Chr.6: 31616953-31616953 on Build GRCh38
rs564950528 Chr.6: 31616934-31616934 on Build GRCh38
rs748101333 Chr.6: 31616834-31616834 on Build GRCh38
rs767750561 Chr.6: 31616930-31616930 on Build GRCh38
rs778733486 Chr.6: 31616839-31616839 on Build GRCh38
rs766411797 Chr.6: 31616831-31616831 on Build GRCh38
rs1032780566 Chr.6: 31616935-31616935 on Build GRCh38
rs368501336 Chr.6: 31616924-31616924 on Build GRCh38
rs746428273 Chr.6: 31616904-31616904 on Build GRCh38
rs764243198 Chr.6: 31616806-31616806 on Build GRCh38
rs148972430 Chr.6: 31616840-31616840 on Build GRCh38
rs775544225 Chr.6: 31616805-31616805 on Build GRCh38
rs771363300 Chr.6: 31616782-31616782 on Build GRCh38
rs764342337 Chr.6: 31616916-31616916 on Build GRCh38
rs746539195 Chr.6: 31616777-31616777 on Build GRCh38
rs753881781 Chr.6: 31616832-31616832 on Build GRCh38
rs766541025 Chr.6: 31616770-31616771 on Build GRCh38
rs775874705 Chr.6: 31616910-31616910 on Build GRCh38
rs765683094 Chr.6: 31616802-31616802 on Build GRCh38
rs141088133 Chr.6: 31616942-31616942 on Build GRCh38
rs777277310 Chr.6: 31616934-31616935 on Build GRCh38
rs187586542 Chr.6: 31616812-31616812 on Build GRCh38
rs770488240 Chr.6: 31616898-31616898 on Build GRCh38
rs36050853 Chr.6: 31616909-31616910 on Build GRCh38
rs777409889 Chr.6: 31616858-31616858 on Build GRCh38
rs145825376 Chr.6: 31616824-31616824 on Build GRCh38
rs757414577 Chr.6: 31616830-31616830 on Build GRCh38
rs777100962 Chr.6: 31616900-31616900 on Build GRCh38
rs1038397741 Chr.6: 31616950-31616950 on Build GRCh38
rs374714373 Chr.6: 31616786-31616786 on Build GRCh38

View Full Product Details