Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179576831-179577092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL6A
Entrez Gene ID
86
Gene Name
actin like 6A
Gene Aliases
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K
UniGene
Hs.435326
Gene Chromosome Location
Chr.3: 179562880-179588405 on Build GRCh38
Transcripts
NM_004301.4 NM_177989.3 NM_178042.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3205856 c.733C>G p.Q245E Chr.3: 179576878-179576878 on Build GRCh38
6326387 c.710G>A p.W237* Chr.3: 179576855-179576855 on Build GRCh38
1421065 c.750G>A p.W250* Chr.3: 179576895-179576895 on Build GRCh38
4980397 c.740C>T p.T247M Chr.3: 179576885-179576885 on Build GRCh38
5026083 c.733C>T p.Q245* Chr.3: 179576878-179576878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752228739 Chr.3: 179576944-179576944 on Build GRCh38
rs572346118 Chr.3: 179576913-179576913 on Build GRCh38
rs754289965 Chr.3: 179576960-179576960 on Build GRCh38
rs769871706 Chr.3: 179576886-179576886 on Build GRCh38
rs919843540 Chr.3: 179576968-179576969 on Build GRCh38
rs577935321 Chr.3: 179576969-179576969 on Build GRCh38
rs767178500 Chr.3: 179576937-179576937 on Build GRCh38
rs563585257 Chr.3: 179576990-179576990 on Build GRCh38
rs543597167 Chr.3: 179576982-179576982 on Build GRCh38
rs753174247 Chr.3: 179576972-179576972 on Build GRCh38
rs2292906 Chr.3: 179577013-179577013 on Build GRCh38
rs754763162 Chr.3: 179576922-179576922 on Build GRCh38
rs979373999 Chr.3: 179577054-179577054 on Build GRCh38
rs780324100 Chr.3: 179576862-179576862 on Build GRCh38
rs773240025 Chr.3: 179576896-179576896 on Build GRCh38
rs868179550 Chr.3: 179576980-179576980 on Build GRCh38
rs2292905 Chr.3: 179577057-179577057 on Build GRCh38
rs559331367 Chr.3: 179577044-179577044 on Build GRCh38
rs781136588 Chr.3: 179576937-179576938 on Build GRCh38
rs762808413 Chr.3: 179576898-179576898 on Build GRCh38
rs1050603697 Chr.3: 179576905-179576905 on Build GRCh38
rs137875597 Chr.3: 179576948-179576948 on Build GRCh38
rs759176888 Chr.3: 179576936-179576936 on Build GRCh38
rs529228372 Chr.3: 179577004-179577004 on Build GRCh38
rs747059728 Chr.3: 179576885-179576885 on Build GRCh38
rs370130128 Chr.3: 179576946-179576946 on Build GRCh38
rs558266605 Chr.3: 179576966-179576966 on Build GRCh38
rs992010798 Chr.3: 179577023-179577023 on Build GRCh38
rs774180938 Chr.3: 179576926-179576926 on Build GRCh38
rs916459784 Chr.3: 179577038-179577038 on Build GRCh38

View Full Product Details