Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49551041-49551274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf141
Entrez Gene ID
135398
Gene Name
chromosome 6 open reading frame 141
Gene Aliases
-
UniGene
Hs.722010
Gene Chromosome Location
Chr.6: 49550400-49560934 on Build GRCh38
Transcripts
NM_001145652.1 XM_005248850.3 XM_005248852.2 XM_005248853.2 XM_006714997.2 XM_011514303.2 XM_017010294.1 XM_017010295.1 XM_017010296.1 XM_017010297.1 XM_017010298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272300 c.334G>T p.G112C Chr.6: 49551126-49551126 on Build GRCh38
1445002 c.401T>A p.F134Y Chr.6: 49551193-49551193 on Build GRCh38
5564480 c.431G>A p.R144H Chr.6: 49551223-49551223 on Build GRCh38
1445001 c.261G>T p.E87D Chr.6: 49551053-49551053 on Build GRCh38
5405504 c.307_308GG>AA p.G103K Chr.6: 49551099-49551100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915375341 Chr.6: 49551187-49551187 on Build GRCh38
rs376050066 Chr.6: 49551105-49551105 on Build GRCh38
rs1049853201 Chr.6: 49551136-49551136 on Build GRCh38
rs191682924 Chr.6: 49551161-49551161 on Build GRCh38
rs989691663 Chr.6: 49551182-49551182 on Build GRCh38
rs112409542 Chr.6: 49551102-49551102 on Build GRCh38
rs372275351 Chr.6: 49551071-49551071 on Build GRCh38
rs776391915 Chr.6: 49551223-49551223 on Build GRCh38
rs773486849 Chr.6: 49551242-49551242 on Build GRCh38
rs575445587 Chr.6: 49551184-49551184 on Build GRCh38
rs766945300 Chr.6: 49551206-49551206 on Build GRCh38
rs768750130 Chr.6: 49551207-49551207 on Build GRCh38
rs865827477 Chr.6: 49551124-49551124 on Build GRCh38
rs748282180 Chr.6: 49551228-49551228 on Build GRCh38
rs6919674 Chr.6: 49551201-49551201 on Build GRCh38
rs760716881 Chr.6: 49551180-49551181 on Build GRCh38
rs756677898 Chr.6: 49551143-49551143 on Build GRCh38
rs763220459 Chr.6: 49551251-49551251 on Build GRCh38
rs773245551 Chr.6: 49551131-49551154 on Build GRCh38
rs896975966 Chr.6: 49551178-49551178 on Build GRCh38
rs976710381 Chr.6: 49551093-49551093 on Build GRCh38
rs965378940 Chr.6: 49551089-49551089 on Build GRCh38
rs1015394271 Chr.6: 49551165-49551165 on Build GRCh38
rs769702589 Chr.6: 49551241-49551241 on Build GRCh38
rs778643506 Chr.6: 49551149-49551149 on Build GRCh38

View Full Product Details