Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 57482103-57482364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTBL2
Entrez Gene ID
345651
Gene Name
actin, beta like 2
Gene Aliases
ACT
UniGene
Hs.482167
Gene Chromosome Location
Chr.5: 57480016-57482809 on Build GRCh38
Transcripts
NM_001017992.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738466 c.389C>A p.T130K Chr.5: 57482319-57482319 on Build GRCh38
5781027 c.570C>G p.L190L Chr.5: 57482138-57482138 on Build GRCh38
5674528 c.392delC p.P131fs*23 Chr.5: 57482316-57482316 on Build GRCh38
3855736 c.521A>G p.H174R Chr.5: 57482187-57482187 on Build GRCh38
1620417 c.404T>C p.V135A Chr.5: 57482304-57482304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551551940 Chr.5: 57482306-57482306 on Build GRCh38
rs566758260 Chr.5: 57482311-57482311 on Build GRCh38
rs777902137 Chr.5: 57482332-57482332 on Build GRCh38
rs779135503 Chr.5: 57482232-57482232 on Build GRCh38
rs199915773 Chr.5: 57482340-57482340 on Build GRCh38
rs150693822 Chr.5: 57482208-57482208 on Build GRCh38
rs755601828 Chr.5: 57482157-57482157 on Build GRCh38
rs1033166885 Chr.5: 57482249-57482249 on Build GRCh38
rs767274326 Chr.5: 57482281-57482281 on Build GRCh38
rs748409406 Chr.5: 57482237-57482237 on Build GRCh38
rs753169150 Chr.5: 57482337-57482337 on Build GRCh38
rs747561123 Chr.5: 57482176-57482176 on Build GRCh38
rs202177504 Chr.5: 57482160-57482160 on Build GRCh38
rs200203178 Chr.5: 57482197-57482197 on Build GRCh38
rs756099192 Chr.5: 57482154-57482154 on Build GRCh38
rs771324969 Chr.5: 57482187-57482187 on Build GRCh38
rs142029713 Chr.5: 57482169-57482169 on Build GRCh38
rs777118554 Chr.5: 57482188-57482188 on Build GRCh38
rs761616119 Chr.5: 57482279-57482279 on Build GRCh38
rs527573101 Chr.5: 57482155-57482155 on Build GRCh38
rs758593298 Chr.5: 57482245-57482245 on Build GRCh38
rs759749382 Chr.5: 57482195-57482195 on Build GRCh38
rs61737336 Chr.5: 57482276-57482276 on Build GRCh38
rs754913467 Chr.5: 57482310-57482310 on Build GRCh38
rs184810267 Chr.5: 57482163-57482163 on Build GRCh38
rs143408227 Chr.5: 57482270-57482270 on Build GRCh38
rs910196893 Chr.5: 57482283-57482283 on Build GRCh38
rs772855283 Chr.5: 57482302-57482302 on Build GRCh38
rs759588880 Chr.5: 57482339-57482339 on Build GRCh38
rs61737335 Chr.5: 57482266-57482266 on Build GRCh38
rs753681748 Chr.5: 57482219-57482219 on Build GRCh38
rs749941418 Chr.5: 57482203-57482203 on Build GRCh38
rs758683068 Chr.5: 57482318-57482318 on Build GRCh38
rs200576412 Chr.5: 57482175-57482175 on Build GRCh38
rs149715690 Chr.5: 57482303-57482303 on Build GRCh38
rs929237089 Chr.5: 57482156-57482156 on Build GRCh38
rs754679525 Chr.5: 57482231-57482231 on Build GRCh38
rs753676520 Chr.5: 57482309-57482309 on Build GRCh38
rs752472175 Chr.5: 57482317-57482317 on Build GRCh38
rs766164059 Chr.5: 57482217-57482217 on Build GRCh38
rs777510878 Chr.5: 57482233-57482234 on Build GRCh38
rs756781690 Chr.5: 57482135-57482135 on Build GRCh38
rs776957958 Chr.5: 57482268-57482268 on Build GRCh38
rs763832753 Chr.5: 57482196-57482196 on Build GRCh38
rs751131522 Chr.5: 57482131-57482131 on Build GRCh38
rs148162647 Chr.5: 57482150-57482150 on Build GRCh38
rs370507258 Chr.5: 57482272-57482272 on Build GRCh38
rs747123764 Chr.5: 57482335-57482335 on Build GRCh38
rs747084615 Chr.5: 57482265-57482265 on Build GRCh38
rs779044293 Chr.5: 57482330-57482341 on Build GRCh38
rs763646977 Chr.5: 57482130-57482130 on Build GRCh38
rs767188513 Chr.5: 57482198-57482198 on Build GRCh38
rs139052385 Chr.5: 57482251-57482251 on Build GRCh38

View Full Product Details