Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35759554-35759821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER3
Entrez Gene ID
148137
Gene Name
proline and serine rich 3
Gene Aliases
C19orf55
UniGene
Hs.527982
Gene Chromosome Location
Chr.19: 35758139-35770115 on Build GRCh38
Transcripts
NM_001039887.2 XM_011526528.1 XM_011526529.2 XM_011526530.1 XM_011526531.1 XM_011526532.1 XM_011526533.2 XM_011526534.1 XM_011526535.2 XM_011526536.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371269 c.132G>C p.Q44H Chr.19: 35759812-35759812 on Build GRCh38
6237770 c.140G>T p.R47M Chr.19: 35759820-35759820 on Build GRCh38
3371271 c.132G>C p.Q44H Chr.19: 35759812-35759812 on Build GRCh38
5508489 c.109-7A>G p.? Chr.19: 35759782-35759782 on Build GRCh38
5508488 c.109-7A>G p.? Chr.19: 35759782-35759782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141430352 Chr.19: 35759670-35759670 on Build GRCh38
rs925465516 Chr.19: 35759776-35759776 on Build GRCh38
rs796384553 Chr.19: 35759655-35759655 on Build GRCh38
rs541874170 Chr.19: 35759679-35759679 on Build GRCh38
rs754579012 Chr.19: 35759773-35759773 on Build GRCh38
rs897076601 Chr.19: 35759604-35759604 on Build GRCh38
rs1017592979 Chr.19: 35759630-35759630 on Build GRCh38
rs1030504032 Chr.19: 35759709-35759709 on Build GRCh38
rs751497875 Chr.19: 35759689-35759689 on Build GRCh38
rs866009234 Chr.19: 35759643-35759643 on Build GRCh38
rs763956553 Chr.19: 35759661-35759661 on Build GRCh38
rs958138696 Chr.19: 35759598-35759598 on Build GRCh38
rs764762152 Chr.19: 35759779-35759779 on Build GRCh38
rs976075009 Chr.19: 35759678-35759678 on Build GRCh38
rs887865989 Chr.19: 35759612-35759612 on Build GRCh38
rs563378666 Chr.19: 35759736-35759736 on Build GRCh38
rs1006741395 Chr.19: 35759617-35759617 on Build GRCh38
rs993968169 Chr.19: 35759607-35759607 on Build GRCh38
rs1046322781 Chr.19: 35759592-35759592 on Build GRCh38
rs911816615 Chr.19: 35759649-35759649 on Build GRCh38
rs966965285 Chr.19: 35759715-35759715 on Build GRCh38
rs936419139 Chr.19: 35759784-35759784 on Build GRCh38
rs987368109 Chr.19: 35759638-35759638 on Build GRCh38
rs989436506 Chr.19: 35759605-35759605 on Build GRCh38
rs7254581 Chr.19: 35759745-35759745 on Build GRCh38
rs762393535 Chr.19: 35759655-35759655 on Build GRCh38
rs753448249 Chr.19: 35759766-35759766 on Build GRCh38
rs1031031726 Chr.19: 35759593-35759593 on Build GRCh38
rs751474207 Chr.19: 35759758-35759759 on Build GRCh38
rs1055559395 Chr.19: 35759753-35759754 on Build GRCh38
rs77797797 Chr.19: 35759610-35759610 on Build GRCh38

View Full Product Details