Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40349669-40349908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSL
Entrez Gene ID
158
Gene Name
adenylosuccinate lyase
Gene Aliases
AMPS, ASASE, ASL
UniGene
Hs.75527
Gene Chromosome Location
Chr.22: 40346500-40387408 on Build GRCh38
Transcripts
NM_000026.3 NM_001123378.2 NM_001317923.1 NR_134256.1 XM_011529977.2 XM_011529979.1 XM_011529980.2 XM_017028636.1 XM_017028637.1 XM_017028638.1 XM_017028639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3992189 c.215T>A p.I72N Chr.22: 40349893-40349893 on Build GRCh38
350322 c.195G>A p.M65I Chr.22: 40349873-40349873 on Build GRCh38
278687 c.229G>A p.A77T Chr.22: 40349907-40349907 on Build GRCh38
4104377 c.190G>A p.E64K Chr.22: 40349868-40349868 on Build GRCh38
273904 c.217G>A p.D73N Chr.22: 40349895-40349895 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756234147 Chr.22: 40349803-40349804 on Build GRCh38
rs765158535 Chr.22: 40349832-40349832 on Build GRCh38
rs752349207 Chr.22: 40349876-40349876 on Build GRCh38
rs544172142 Chr.22: 40349851-40349851 on Build GRCh38
rs947666693 Chr.22: 40349762-40349762 on Build GRCh38
rs764558921 Chr.22: 40349803-40349803 on Build GRCh38
rs762257005 Chr.22: 40349823-40349823 on Build GRCh38
rs760976951 Chr.22: 40349800-40349800 on Build GRCh38
rs983022020 Chr.22: 40349714-40349714 on Build GRCh38
rs750871310 Chr.22: 40349847-40349847 on Build GRCh38
rs767005918 Chr.22: 40349867-40349867 on Build GRCh38
rs558339570 Chr.22: 40349718-40349718 on Build GRCh38
rs750583169 Chr.22: 40349789-40349790 on Build GRCh38
rs576810607 Chr.22: 40349768-40349768 on Build GRCh38
rs200169254 Chr.22: 40349883-40349883 on Build GRCh38
rs962889414 Chr.22: 40349764-40349764 on Build GRCh38
rs777004146 Chr.22: 40349817-40349817 on Build GRCh38
rs974269660 Chr.22: 40349766-40349766 on Build GRCh38
rs772257987 Chr.22: 40349789-40349789 on Build GRCh38
rs796052251 Chr.22: 40349875-40349875 on Build GRCh38
rs775914684 Chr.22: 40349796-40349796 on Build GRCh38
rs765813625 Chr.22: 40349839-40349839 on Build GRCh38
rs917591459 Chr.22: 40349740-40349740 on Build GRCh38

View Full Product Details