Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67291810-67292051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13D
Entrez Gene ID
338692
Gene Name
ankyrin repeat domain 13D
Gene Aliases
-
UniGene
Hs.438673
Gene Chromosome Location
Chr.11: 67289291-67302484 on Build GRCh38
Transcripts
NM_207354.2 NR_030767.1 XM_011544985.1 XM_011544986.1 XM_011544987.2 XM_011544988.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
690376 c.323T>G p.V108G Chr.11: 67292033-67292033 on Build GRCh38
6418894 c.592G>T p.V198L Chr.11: 67292041-67292041 on Build GRCh38
5619038 c.579G>T p.R193R Chr.11: 67292028-67292028 on Build GRCh38
6392499 c.581A>G p.Q194R Chr.11: 67292030-67292030 on Build GRCh38
2041490 c.595G>T p.E199* Chr.11: 67292044-67292044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751959550 Chr.11: 67291942-67291942 on Build GRCh38
rs201999693 Chr.11: 67292031-67292031 on Build GRCh38
rs748581543 Chr.11: 67291922-67291922 on Build GRCh38
rs776371000 Chr.11: 67291980-67291980 on Build GRCh38
rs1033534969 Chr.11: 67291893-67291893 on Build GRCh38
rs564526871 Chr.11: 67291894-67291894 on Build GRCh38
rs1021000582 Chr.11: 67291835-67291835 on Build GRCh38
rs764845999 Chr.11: 67291985-67291985 on Build GRCh38
rs532012832 Chr.11: 67291904-67291904 on Build GRCh38
rs376543024 Chr.11: 67291986-67291986 on Build GRCh38
rs757711478 Chr.11: 67291999-67291999 on Build GRCh38
rs970678075 Chr.11: 67291856-67291856 on Build GRCh38
rs546133728 Chr.11: 67291858-67291858 on Build GRCh38
rs35965170 Chr.11: 67292028-67292028 on Build GRCh38
rs909294424 Chr.11: 67291921-67291921 on Build GRCh38
rs566719888 Chr.11: 67292002-67292002 on Build GRCh38
rs746799045 Chr.11: 67291977-67291977 on Build GRCh38
rs939258655 Chr.11: 67291938-67291938 on Build GRCh38
rs138461637 Chr.11: 67292025-67292025 on Build GRCh38
rs750688587 Chr.11: 67292026-67292026 on Build GRCh38
rs550381557 Chr.11: 67291925-67291925 on Build GRCh38
rs371802130 Chr.11: 67292027-67292027 on Build GRCh38
rs915301913 Chr.11: 67291903-67291903 on Build GRCh38
rs959181733 Chr.11: 67291902-67291902 on Build GRCh38
rs1008204486 Chr.11: 67291932-67291947 on Build GRCh38
rs752047469 Chr.11: 67291993-67291993 on Build GRCh38
rs771970484 Chr.11: 67291972-67291972 on Build GRCh38
rs773045418 Chr.11: 67291975-67291975 on Build GRCh38
rs373465801 Chr.11: 67291982-67291982 on Build GRCh38
rs369287312 Chr.11: 67291943-67291943 on Build GRCh38
rs972100686 Chr.11: 67291923-67291923 on Build GRCh38
rs991606778 Chr.11: 67291918-67291918 on Build GRCh38
rs748157475 Chr.11: 67291964-67291964 on Build GRCh38
rs529766998 Chr.11: 67291959-67291959 on Build GRCh38
rs779956748 Chr.11: 67291962-67291962 on Build GRCh38
rs764693875 Chr.11: 67291992-67291992 on Build GRCh38
rs932780333 Chr.11: 67291833-67291833 on Build GRCh38
rs746016813 Chr.11: 67291960-67291960 on Build GRCh38

View Full Product Details