Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136679460-136679729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF6
Entrez Gene ID
9459
Gene Name
Rac/Cdc42 guanine nucleotide exchange factor 6
Gene Aliases
COOL2, Cool-2, MRX46, PIXA, alpha-PIX, alphaPIX
UniGene
Hs.522795
Gene Chromosome Location
Chr.X: 136665547-136781656 on Build GRCh38
Transcripts
NM_001306177.1 NM_004840.2 XM_005262499.2 XM_011531412.2 XM_011531413.1 XM_011531414.1 XM_011531415.2 XM_011531416.2 XM_011531417.1 XM_017029975.1 XM_017029976.1

SNP Detail

SNP ID Chromosome Location
rs776423256 Chr.X: 136679619-136679619 on Build GRCh38
rs773106139 Chr.X: 136679585-136679585 on Build GRCh38
rs749344427 Chr.X: 136679709-136679709 on Build GRCh38
rs780063494 Chr.X: 136679559-136679559 on Build GRCh38
rs371381075 Chr.X: 136679496-136679496 on Build GRCh38
rs751849274 Chr.X: 136679519-136679519 on Build GRCh38
rs775098750 Chr.X: 136679635-136679635 on Build GRCh38
rs369009271 Chr.X: 136679494-136679494 on Build GRCh38
rs770679754 Chr.X: 136679602-136679602 on Build GRCh38
rs768051577 Chr.X: 136679627-136679627 on Build GRCh38
rs757559045 Chr.X: 136679521-136679521 on Build GRCh38
rs1009763711 Chr.X: 136679697-136679697 on Build GRCh38
rs763553724 Chr.X: 136679638-136679638 on Build GRCh38
rs148319648 Chr.X: 136679646-136679646 on Build GRCh38
rs771876538 Chr.X: 136679579-136679579 on Build GRCh38
rs375053756 Chr.X: 136679526-136679526 on Build GRCh38
rs750669987 Chr.X: 136679700-136679700 on Build GRCh38
rs1054018399 Chr.X: 136679683-136679683 on Build GRCh38
rs764498295 Chr.X: 136679639-136679639 on Build GRCh38
rs369167264 Chr.X: 136679706-136679706 on Build GRCh38
rs374950341 Chr.X: 136679488-136679488 on Build GRCh38
rs917669569 Chr.X: 136679533-136679533 on Build GRCh38
rs759207016 Chr.X: 136679624-136679624 on Build GRCh38
rs761281525 Chr.X: 136679707-136679707 on Build GRCh38
rs781267386 Chr.X: 136679522-136679522 on Build GRCh38
rs145081421 Chr.X: 136679601-136679601 on Build GRCh38
rs749749273 Chr.X: 136679694-136679694 on Build GRCh38
rs938254089 Chr.X: 136679597-136679597 on Build GRCh38
rs936920662 Chr.X: 136679650-136679650 on Build GRCh38
rs748119122 Chr.X: 136679573-136679573 on Build GRCh38
rs926622130 Chr.X: 136679552-136679552 on Build GRCh38
rs756204695 Chr.X: 136679539-136679539 on Build GRCh38

View Full Product Details