Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 6887076-6887329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP28
Entrez Gene ID
79822
Gene Name
Rho GTPase activating protein 28
Gene Aliases
-
UniGene
Hs.183114
Gene Chromosome Location
Chr.18: 6729718-6915716 on Build GRCh38
Transcripts
NM_001010000.2 XM_005258144.1 XM_005258145.1 XM_005258146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349491 c.985C>G p.H329D Chr.18: 6887165-6887165 on Build GRCh38
3692232 c.1012C>T p.H338Y Chr.18: 6887192-6887192 on Build GRCh38
7298545 c.1459C>T p.P487S Chr.18: 6887162-6887162 on Build GRCh38
3145658 c.1536+5G>A p.? Chr.18: 6887244-6887244 on Build GRCh38
4940314 c.1464C>T p.H488H Chr.18: 6887167-6887167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754953104 Chr.18: 6887276-6887276 on Build GRCh38
rs760753313 Chr.18: 6887229-6887229 on Build GRCh38
rs777432146 Chr.18: 6887131-6887131 on Build GRCh38
rs768612992 Chr.18: 6887204-6887204 on Build GRCh38
rs148642656 Chr.18: 6887224-6887224 on Build GRCh38
rs779401872 Chr.18: 6887272-6887272 on Build GRCh38
rs776248494 Chr.18: 6887219-6887220 on Build GRCh38
rs372564720 Chr.18: 6887244-6887244 on Build GRCh38
rs766815321 Chr.18: 6887243-6887243 on Build GRCh38
rs993358693 Chr.18: 6887293-6887293 on Build GRCh38
rs749874144 Chr.18: 6887227-6887228 on Build GRCh38
rs933395202 Chr.18: 6887135-6887135 on Build GRCh38
rs762843575 Chr.18: 6887134-6887134 on Build GRCh38
rs774266585 Chr.18: 6887208-6887208 on Build GRCh38
rs370652388 Chr.18: 6887209-6887209 on Build GRCh38
rs147455581 Chr.18: 6887128-6887128 on Build GRCh38
rs545031750 Chr.18: 6887267-6887267 on Build GRCh38
rs746610138 Chr.18: 6887119-6887119 on Build GRCh38
rs368842280 Chr.18: 6887228-6887228 on Build GRCh38
rs559381938 Chr.18: 6887122-6887122 on Build GRCh38
rs374741702 Chr.18: 6887248-6887248 on Build GRCh38
rs374782517 Chr.18: 6887161-6887161 on Build GRCh38
rs745378361 Chr.18: 6887167-6887167 on Build GRCh38
rs878867026 Chr.18: 6887217-6887217 on Build GRCh38
rs751248905 Chr.18: 6887142-6887142 on Build GRCh38
rs767129529 Chr.18: 6887251-6887251 on Build GRCh38
rs747888905 Chr.18: 6887289-6887289 on Build GRCh38
rs935993270 Chr.18: 6887223-6887223 on Build GRCh38
rs749070329 Chr.18: 6887168-6887168 on Build GRCh38
rs376217018 Chr.18: 6887246-6887246 on Build GRCh38
rs749912613 Chr.18: 6887257-6887257 on Build GRCh38
rs778922194 Chr.18: 6887287-6887287 on Build GRCh38
rs564990083 Chr.18: 6887249-6887249 on Build GRCh38
rs765634994 Chr.18: 6887247-6887247 on Build GRCh38
rs768329258 Chr.18: 6887214-6887215 on Build GRCh38
rs559048725 Chr.18: 6887121-6887121 on Build GRCh38
rs756876902 Chr.18: 6887155-6887155 on Build GRCh38
rs533552294 Chr.18: 6887259-6887259 on Build GRCh38
rs761155628 Chr.18: 6887107-6887107 on Build GRCh38
rs369277583 Chr.18: 6887262-6887262 on Build GRCh38
rs925509507 Chr.18: 6887095-6887095 on Build GRCh38

View Full Product Details