Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151116580-151116853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP3
Entrez Gene ID
116988
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3
UniGene
Hs.647075
Gene Chromosome Location
Chr.7: 151085831-151144436 on Build GRCh38
Transcripts
NM_001042535.3 NM_001281300.1 NM_001308304.1 NM_001308305.1 NM_031946.6 XM_005249943.2 XM_011515780.1 XM_017011732.1 XM_017011733.1 XM_017011734.1 XM_017011735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7141485 c.342G>T p.V114V Chr.7: 151116803-151116803 on Build GRCh38
6596061 c.378G>A p.P126P Chr.7: 151116839-151116839 on Build GRCh38
6596080 c.336G>A p.S112S Chr.7: 151116797-151116797 on Build GRCh38
1488374 c.335C>T p.S112L Chr.7: 151116796-151116796 on Build GRCh38
6596079 c.336G>A p.S112S Chr.7: 151116797-151116797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051543641 Chr.7: 151116708-151116708 on Build GRCh38
rs1038444729 Chr.7: 151116669-151116669 on Build GRCh38
rs1007272346 Chr.7: 151116634-151116634 on Build GRCh38
rs769561471 Chr.7: 151116780-151116780 on Build GRCh38
rs375303035 Chr.7: 151116770-151116770 on Build GRCh38
rs755096478 Chr.7: 151116828-151116828 on Build GRCh38
rs748402014 Chr.7: 151116744-151116744 on Build GRCh38
rs766557940 Chr.7: 151116820-151116820 on Build GRCh38
rs371042682 Chr.7: 151116788-151116788 on Build GRCh38
rs73169605 Chr.7: 151116668-151116668 on Build GRCh38
rs1048712730 Chr.7: 151116615-151116615 on Build GRCh38
rs890345193 Chr.7: 151116709-151116709 on Build GRCh38
rs558776775 Chr.7: 151116646-151116646 on Build GRCh38
rs779219909 Chr.7: 151116829-151116829 on Build GRCh38
rs1010945269 Chr.7: 151116729-151116729 on Build GRCh38
rs759457994 Chr.7: 151116775-151116775 on Build GRCh38
rs368165513 Chr.7: 151116771-151116771 on Build GRCh38
rs776190317 Chr.7: 151116774-151116774 on Build GRCh38
rs750655059 Chr.7: 151116806-151116806 on Build GRCh38
rs753276868 Chr.7: 151116831-151116831 on Build GRCh38
rs560531453 Chr.7: 151116676-151116676 on Build GRCh38
rs192957763 Chr.7: 151116789-151116789 on Build GRCh38
rs370623093 Chr.7: 151116745-151116745 on Build GRCh38
rs527444975 Chr.7: 151116645-151116645 on Build GRCh38
rs552474810 Chr.7: 151116688-151116688 on Build GRCh38
rs761014833 Chr.7: 151116812-151116812 on Build GRCh38
rs1040061813 Chr.7: 151116664-151116664 on Build GRCh38
rs548334424 Chr.7: 151116797-151116797 on Build GRCh38
rs980868438 Chr.7: 151116772-151116772 on Build GRCh38
rs932632736 Chr.7: 151116701-151116701 on Build GRCh38
rs777250594 Chr.7: 151116758-151116758 on Build GRCh38
rs532633679 Chr.7: 151116686-151116686 on Build GRCh38
rs146303814 Chr.7: 151116821-151116821 on Build GRCh38
rs1052326765 Chr.7: 151116748-151116748 on Build GRCh38
rs561814511 Chr.7: 151116629-151116629 on Build GRCh38
rs778199777 Chr.7: 151116707-151116707 on Build GRCh38
rs772487965 Chr.7: 151116755-151116755 on Build GRCh38
rs957307356 Chr.7: 151116728-151116728 on Build GRCh38
rs368600470 Chr.7: 151116833-151116833 on Build GRCh38
rs369739515 Chr.7: 151116753-151116753 on Build GRCh38

View Full Product Details