Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 60351655-60351900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA2
Entrez Gene ID
302
Gene Name
annexin A2
Gene Aliases
ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
UniGene
Hs.511605
Gene Chromosome Location
Chr.15: 60347151-60398025 on Build GRCh38
Transcripts
NM_001002857.1 NM_001002858.2 NM_001136015.2 NM_004039.2 XM_011521477.2 XM_017022090.1 XM_017022091.1 XM_017022092.1 XM_017022093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608525 c.777G>A p.L259L Chr.15: 60351779-60351779 on Build GRCh38
3377605 c.742G>C p.D248H Chr.15: 60351814-60351814 on Build GRCh38
4747347 c.815delA p.N272fs*4 Chr.15: 60351741-60351741 on Build GRCh38
3711836 c.822C>T p.F274F Chr.15: 60351734-60351734 on Build GRCh38
5383618 c.808C>T p.L270L Chr.15: 60351748-60351748 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762224831 Chr.15: 60351867-60351867 on Build GRCh38
rs930238328 Chr.15: 60351709-60351709 on Build GRCh38
rs780280219 Chr.15: 60351769-60351769 on Build GRCh38
rs773163394 Chr.15: 60351848-60351848 on Build GRCh38
rs879201142 Chr.15: 60351802-60351802 on Build GRCh38
rs140356576 Chr.15: 60351812-60351812 on Build GRCh38
rs11633619 Chr.15: 60351875-60351875 on Build GRCh38
rs755187006 Chr.15: 60351844-60351844 on Build GRCh38
rs749930415 Chr.15: 60351706-60351706 on Build GRCh38
rs750920398 Chr.15: 60351831-60351831 on Build GRCh38
rs776413720 Chr.15: 60351782-60351782 on Build GRCh38
rs761751271 Chr.15: 60351686-60351686 on Build GRCh38
rs766033415 Chr.15: 60351720-60351720 on Build GRCh38
rs747214524 Chr.15: 60351778-60351778 on Build GRCh38
rs779634258 Chr.15: 60351758-60351758 on Build GRCh38
rs749207144 Chr.15: 60351849-60351849 on Build GRCh38
rs368401421 Chr.15: 60351717-60351717 on Build GRCh38
rs192656325 Chr.15: 60351799-60351799 on Build GRCh38
rs757758309 Chr.15: 60351816-60351816 on Build GRCh38
rs376757847 Chr.15: 60351779-60351779 on Build GRCh38
rs776498936 Chr.15: 60351678-60351678 on Build GRCh38
rs774007320 Chr.15: 60351806-60351806 on Build GRCh38
rs765805060 Chr.15: 60351825-60351825 on Build GRCh38
rs371940535 Chr.15: 60351784-60351784 on Build GRCh38
rs758443986 Chr.15: 60351768-60351768 on Build GRCh38
rs145435452 Chr.15: 60351813-60351813 on Build GRCh38
rs769618158 Chr.15: 60351847-60351847 on Build GRCh38
rs185041670 Chr.15: 60351851-60351851 on Build GRCh38
rs775318031 Chr.15: 60351852-60351852 on Build GRCh38
rs745934073 Chr.15: 60351764-60351764 on Build GRCh38
rs747188095 Chr.15: 60351843-60351843 on Build GRCh38
rs376822302 Chr.15: 60351811-60351811 on Build GRCh38
rs754340964 Chr.15: 60351739-60351739 on Build GRCh38
rs112023664 Chr.15: 60351723-60351723 on Build GRCh38
rs756463278 Chr.15: 60351727-60351727 on Build GRCh38
rs116283617 Chr.15: 60351731-60351731 on Build GRCh38
rs780008022 Chr.15: 60351840-60351840 on Build GRCh38
rs769705842 Chr.15: 60351788-60351788 on Build GRCh38
rs372199283 Chr.15: 60351725-60351725 on Build GRCh38
rs768365129 Chr.15: 60351677-60351677 on Build GRCh38
rs761886498 Chr.15: 60351728-60351728 on Build GRCh38
rs757848231 Chr.15: 60351753-60351753 on Build GRCh38
rs758452893 Chr.15: 60351839-60351839 on Build GRCh38
rs747916256 Chr.15: 60351845-60351845 on Build GRCh38

View Full Product Details