Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77199754-77200003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5411314 c.110C>T p.A37V Chr.9: 77199954-77199954 on Build GRCh38
5935529 c.144+1G>A p.? Chr.9: 77199989-77199989 on Build GRCh38
4740404 c.117G>T p.K39N Chr.9: 77199961-77199961 on Build GRCh38
6820154 c.143T>C p.L48P Chr.9: 77199987-77199987 on Build GRCh38
5411315 c.110C>T p.A37V Chr.9: 77199954-77199954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918572315 Chr.9: 77199790-77199790 on Build GRCh38
rs990061243 Chr.9: 77199928-77199928 on Build GRCh38
rs202024224 Chr.9: 77199931-77199931 on Build GRCh38
rs762167465 Chr.9: 77199941-77199941 on Build GRCh38
rs769097566 Chr.9: 77199936-77199936 on Build GRCh38
rs960135788 Chr.9: 77199960-77199960 on Build GRCh38
rs758443738 Chr.9: 77199936-77199937 on Build GRCh38
rs372018749 Chr.9: 77199973-77199973 on Build GRCh38
rs527950248 Chr.9: 77199870-77199870 on Build GRCh38
rs752636187 Chr.9: 77199937-77199937 on Build GRCh38
rs747022718 Chr.9: 77199799-77199801 on Build GRCh38
rs140518771 Chr.9: 77199970-77199970 on Build GRCh38
rs368914910 Chr.9: 77199962-77199962 on Build GRCh38
rs1051397652 Chr.9: 77199922-77199922 on Build GRCh38
rs933055967 Chr.9: 77199908-77199908 on Build GRCh38
rs764893746 Chr.9: 77199925-77199925 on Build GRCh38
rs1013695508 Chr.9: 77199797-77199797 on Build GRCh38
rs185381432 Chr.9: 77199839-77199839 on Build GRCh38
rs773656342 Chr.9: 77199968-77199968 on Build GRCh38
rs895343909 Chr.9: 77199796-77199796 on Build GRCh38
rs770319746 Chr.9: 77199947-77199947 on Build GRCh38
rs535834952 Chr.9: 77199882-77199882 on Build GRCh38
rs1031301708 Chr.9: 77199888-77199888 on Build GRCh38
rs971120184 Chr.9: 77199881-77199881 on Build GRCh38
rs957530886 Chr.9: 77199916-77199916 on Build GRCh38
rs1043664164 Chr.9: 77199932-77199932 on Build GRCh38
rs190684534 Chr.9: 77199938-77199938 on Build GRCh38
rs749639590 Chr.9: 77199845-77199848 on Build GRCh38
rs12552898 Chr.9: 77199883-77199883 on Build GRCh38

View Full Product Details