Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32494980-32495241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1C
Entrez Gene ID
59271
Gene Name
eva-1 homolog C (C. elegans)
Gene Aliases
B18, B19, C21orf63, C21orf64, FAM176C, PRED34, SUE21
UniGene
Hs.208358
Gene Chromosome Location
Chr.21: 32411692-32515400 on Build GRCh38
Transcripts
NM_001286556.1 NM_001320744.1 NM_001320745.1 NM_058187.4 NR_104472.1 NR_135467.1 XM_005261019.4 XM_006724038.3 XM_011529669.2 XM_017028418.1 XM_017028419.1 XM_017028420.1 XM_017028421.1 XM_017028422.1 XM_017028423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682738 c.713A>G p.N238S Chr.21: 32495105-32495105 on Build GRCh38
3550269 c.644C>T p.S215F Chr.21: 32495036-32495036 on Build GRCh38
6446754 c.695G>A p.C232Y Chr.21: 32495087-32495087 on Build GRCh38
6533206 c.672A>C p.R224R Chr.21: 32495064-32495064 on Build GRCh38
5959391 c.715C>T p.H239Y Chr.21: 32495107-32495107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762283599 Chr.21: 32495063-32495063 on Build GRCh38
rs774090293 Chr.21: 32495049-32495049 on Build GRCh38
rs748073128 Chr.21: 32495011-32495011 on Build GRCh38
rs755252652 Chr.21: 32495196-32495196 on Build GRCh38
rs761143906 Chr.21: 32495054-32495054 on Build GRCh38
rs2276222 Chr.21: 32495218-32495218 on Build GRCh38
rs754993684 Chr.21: 32495185-32495186 on Build GRCh38
rs762523937 Chr.21: 32495038-32495038 on Build GRCh38
rs749067113 Chr.21: 32495140-32495140 on Build GRCh38
rs765768777 Chr.21: 32495067-32495067 on Build GRCh38
rs146299638 Chr.21: 32495060-32495060 on Build GRCh38
rs149253304 Chr.21: 32495154-32495154 on Build GRCh38
rs61739732 Chr.21: 32495163-32495163 on Build GRCh38
rs777597584 Chr.21: 32495213-32495213 on Build GRCh38
rs200775525 Chr.21: 32495136-32495136 on Build GRCh38
rs750893143 Chr.21: 32495182-32495182 on Build GRCh38
rs139276987 Chr.21: 32495098-32495098 on Build GRCh38
rs780272765 Chr.21: 32495086-32495086 on Build GRCh38
rs145839776 Chr.21: 32495009-32495009 on Build GRCh38
rs779730010 Chr.21: 32495096-32495098 on Build GRCh38
rs770419820 Chr.21: 32495036-32495036 on Build GRCh38
rs751614975 Chr.21: 32495187-32495187 on Build GRCh38
rs923796806 Chr.21: 32495097-32495097 on Build GRCh38
rs112199125 Chr.21: 32495212-32495212 on Build GRCh38
rs769690328 Chr.21: 32495124-32495124 on Build GRCh38
rs755880935 Chr.21: 32495209-32495209 on Build GRCh38
rs749212905 Chr.21: 32495215-32495215 on Build GRCh38
rs199942564 Chr.21: 32495205-32495205 on Build GRCh38
rs994978510 Chr.21: 32495217-32495217 on Build GRCh38
rs778960555 Chr.21: 32495216-32495217 on Build GRCh38
rs375469757 Chr.21: 32495062-32495062 on Build GRCh38
rs768388099 Chr.21: 32495002-32495002 on Build GRCh38
rs775994136 Chr.21: 32495085-32495085 on Build GRCh38
rs762168067 Chr.21: 32495166-32495166 on Build GRCh38
rs62214671 Chr.21: 32495071-32495071 on Build GRCh38
rs770516737 Chr.21: 32495141-32495141 on Build GRCh38
rs565328798 Chr.21: 32495164-32495164 on Build GRCh38
rs751737735 Chr.21: 32495096-32495096 on Build GRCh38
rs61744983 Chr.21: 32495137-32495137 on Build GRCh38
rs770716681 Chr.21: 32495045-32495045 on Build GRCh38
rs769751491 Chr.21: 32495015-32495015 on Build GRCh38
rs2276221 Chr.21: 32495186-32495186 on Build GRCh38
rs572171920 Chr.21: 32495099-32495099 on Build GRCh38
rs144435629 Chr.21: 32495204-32495204 on Build GRCh38
rs377141391 Chr.21: 32495192-32495192 on Build GRCh38
rs747965394 Chr.21: 32495105-32495105 on Build GRCh38
rs763051250 Chr.21: 32495183-32495183 on Build GRCh38
rs921527710 Chr.21: 32495050-32495050 on Build GRCh38
rs759305143 Chr.21: 32495160-32495160 on Build GRCh38

View Full Product Details