Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70295953-70296226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
UniGene
Hs.744989
Gene Chromosome Location
Chr.1: 70259002-70354977 on Build GRCh38
Transcripts
NM_030816.4 XM_005271234.3 XM_005271235.3 XM_006710929.3 XM_017002413.1 XM_017002414.1 XM_017002415.1 XM_017002416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2240939 c.960A>G p.L320L Chr.1: 70296223-70296223 on Build GRCh38
4427607 c.1013C>G p.T338R Chr.1: 70296170-70296170 on Build GRCh38
6600382 c.1039C>T p.R347W Chr.1: 70296144-70296144 on Build GRCh38
4773924 c.997A>C p.K333Q Chr.1: 70296186-70296186 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs150473953 Chr.1: 70296130-70296130 on Build GRCh38
rs376778443 Chr.1: 70296088-70296088 on Build GRCh38
rs959705941 Chr.1: 70296075-70296075 on Build GRCh38
rs567155615 Chr.1: 70295979-70295979 on Build GRCh38
rs760688301 Chr.1: 70296085-70296085 on Build GRCh38
rs1035068528 Chr.1: 70296066-70296066 on Build GRCh38
rs1003947375 Chr.1: 70296056-70296056 on Build GRCh38
rs966943929 Chr.1: 70296048-70296048 on Build GRCh38
rs368071100 Chr.1: 70296150-70296150 on Build GRCh38
rs893948262 Chr.1: 70295983-70295983 on Build GRCh38
rs983962408 Chr.1: 70296140-70296140 on Build GRCh38
rs1004068391 Chr.1: 70296062-70296062 on Build GRCh38
rs778182656 Chr.1: 70296169-70296169 on Build GRCh38
rs772209580 Chr.1: 70296079-70296079 on Build GRCh38
rs773096570 Chr.1: 70296085-70296089 on Build GRCh38
rs200906843 Chr.1: 70296090-70296090 on Build GRCh38
rs749959800 Chr.1: 70296102-70296102 on Build GRCh38
rs372460619 Chr.1: 70296089-70296089 on Build GRCh38
rs952483250 Chr.1: 70296131-70296131 on Build GRCh38
rs1020942628 Chr.1: 70296018-70296018 on Build GRCh38
rs772578514 Chr.1: 70296168-70296168 on Build GRCh38
rs76489884 Chr.1: 70296028-70296028 on Build GRCh38
rs779248014 Chr.1: 70296165-70296165 on Build GRCh38
rs371252132 Chr.1: 70296181-70296181 on Build GRCh38
rs574263813 Chr.1: 70296009-70296009 on Build GRCh38
rs61782668 Chr.1: 70296039-70296039 on Build GRCh38
rs145127266 Chr.1: 70296189-70296189 on Build GRCh38
rs773270120 Chr.1: 70296081-70296081 on Build GRCh38
rs34940869 Chr.1: 70296143-70296144 on Build GRCh38
rs748714263 Chr.1: 70296167-70296167 on Build GRCh38
rs375304141 Chr.1: 70295990-70295990 on Build GRCh38
rs747085829 Chr.1: 70296170-70296170 on Build GRCh38
rs753511738 Chr.1: 70296158-70296158 on Build GRCh38
rs769748358 Chr.1: 70296083-70296083 on Build GRCh38
rs767507008 Chr.1: 70296097-70296097 on Build GRCh38
rs770990181 Chr.1: 70296180-70296180 on Build GRCh38

View Full Product Details