Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 134527861-134528098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKR1B10
Entrez Gene ID
57016
Gene Name
aldo-keto reductase family 1 member B10
Gene Aliases
AKR1B11, AKR1B12, ALDRLn, ARL-1, ARL1, HIS, HSI
UniGene
Hs.116724
Gene Chromosome Location
Chr.7: 134527592-134541414 on Build GRCh38
Transcripts
NM_020299.4 XM_011516416.1 XM_011516417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3928831 c.42C>T p.P14P Chr.7: 134527953-134527953 on Build GRCh38
1755093 c.34A>T p.K12* Chr.7: 134527945-134527945 on Build GRCh38
5750211 c.5C>A p.A2D Chr.7: 134527916-134527916 on Build GRCh38
3411601 c.8C>T p.T3M Chr.7: 134527919-134527919 on Build GRCh38
4660906 c.9G>A p.T3T Chr.7: 134527920-134527920 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754091937 Chr.7: 134527937-134527937 on Build GRCh38
rs149691678 Chr.7: 134527950-134527950 on Build GRCh38
rs751410564 Chr.7: 134528044-134528044 on Build GRCh38
rs2005412 Chr.7: 134527920-134527920 on Build GRCh38
rs139571012 Chr.7: 134527938-134527938 on Build GRCh38
rs370608251 Chr.7: 134528021-134528021 on Build GRCh38
rs755703365 Chr.7: 134527982-134527982 on Build GRCh38
rs941140625 Chr.7: 134528069-134528069 on Build GRCh38
rs779810370 Chr.7: 134527983-134527983 on Build GRCh38
rs755191438 Chr.7: 134527948-134527948 on Build GRCh38
rs757140278 Chr.7: 134527953-134527953 on Build GRCh38
rs756835627 Chr.7: 134528076-134528076 on Build GRCh38
rs781283353 Chr.7: 134527954-134527954 on Build GRCh38
rs200830270 Chr.7: 134527999-134527999 on Build GRCh38
rs2037005 Chr.7: 134527989-134527989 on Build GRCh38
rs765565981 Chr.7: 134528026-134528026 on Build GRCh38
rs779132332 Chr.7: 134527997-134527997 on Build GRCh38
rs772692413 Chr.7: 134527922-134527922 on Build GRCh38
rs11545284 Chr.7: 134527932-134527932 on Build GRCh38
rs572830188 Chr.7: 134527930-134527930 on Build GRCh38
rs760757911 Chr.7: 134527929-134527929 on Build GRCh38
rs748859754 Chr.7: 134527987-134527987 on Build GRCh38
rs774817445 Chr.7: 134527916-134527916 on Build GRCh38
rs770968813 Chr.7: 134528016-134528016 on Build GRCh38
rs749448464 Chr.7: 134527905-134527906 on Build GRCh38
rs919472968 Chr.7: 134528000-134528000 on Build GRCh38
rs888212711 Chr.7: 134528068-134528068 on Build GRCh38
rs779005371 Chr.7: 134528003-134528005 on Build GRCh38
rs773276163 Chr.7: 134527927-134527927 on Build GRCh38
rs576663057 Chr.7: 134528070-134528070 on Build GRCh38
rs768931250 Chr.7: 134527883-134527883 on Build GRCh38
rs141459829 Chr.7: 134528001-134528001 on Build GRCh38
rs371340786 Chr.7: 134528020-134528020 on Build GRCh38
rs749890611 Chr.7: 134527977-134527977 on Build GRCh38
rs370374498 Chr.7: 134527962-134527962 on Build GRCh38
rs556642669 Chr.7: 134528066-134528066 on Build GRCh38
rs773730938 Chr.7: 134528003-134528003 on Build GRCh38

View Full Product Details