Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6824971-6825224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
UniGene
Hs.397705
Gene Chromosome Location
Chr.1: 6785324-7769706 on Build GRCh38
Transcripts
NM_001195563.1 NM_001242701.1 NM_015215.3 NR_038934.1 XM_011541083.2 XM_011541084.2 XM_011541085.2 XM_011541086.2 XM_011541087.2 XM_011541088.2 XM_011541089.2 XM_011541090.2 XM_011541091.2 XM_011541092.2 XM_017000773.1 XM_017000774.1 XM_017000775.1 XM_017000776.1 XM_017000777.1 XM_017000778.1 XM_017000779.1 XM_017000780.1 XM_017000781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6540045 c.213G>T p.R71S Chr.1: 6825189-6825189 on Build GRCh38
414800 c.153C>G p.I51M Chr.1: 6825129-6825129 on Build GRCh38
6506882 c.212G>A p.R71K Chr.1: 6825188-6825188 on Build GRCh38
7411316 c.153C>A p.I51I Chr.1: 6825129-6825129 on Build GRCh38
6540046 c.213G>T p.R71S Chr.1: 6825189-6825189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980338158 Chr.1: 6825087-6825087 on Build GRCh38
rs763307751 Chr.1: 6825150-6825150 on Build GRCh38
rs769016348 Chr.1: 6825159-6825159 on Build GRCh38
rs141738594 Chr.1: 6825174-6825174 on Build GRCh38
rs781069182 Chr.1: 6825105-6825105 on Build GRCh38
rs750328495 Chr.1: 6825051-6825051 on Build GRCh38
rs12138800 Chr.1: 6825014-6825014 on Build GRCh38
rs1027741830 Chr.1: 6825010-6825010 on Build GRCh38
rs753277018 Chr.1: 6825047-6825047 on Build GRCh38
rs370617347 Chr.1: 6825109-6825109 on Build GRCh38
rs753668929 Chr.1: 6825061-6825061 on Build GRCh38
rs374088514 Chr.1: 6825110-6825110 on Build GRCh38
rs367805401 Chr.1: 6825121-6825121 on Build GRCh38
rs761250458 Chr.1: 6825048-6825048 on Build GRCh38
rs749875732 Chr.1: 6825164-6825164 on Build GRCh38
rs766218807 Chr.1: 6825177-6825177 on Build GRCh38
rs1005367781 Chr.1: 6825020-6825020 on Build GRCh38
rs34440382 Chr.1: 6825139-6825139 on Build GRCh38
rs770180119 Chr.1: 6825120-6825120 on Build GRCh38
rs535251766 Chr.1: 6825057-6825057 on Build GRCh38
rs912695893 Chr.1: 6825035-6825035 on Build GRCh38
rs879255532 Chr.1: 6825130-6825130 on Build GRCh38
rs775828172 Chr.1: 6825070-6825070 on Build GRCh38
rs746000883 Chr.1: 6825119-6825119 on Build GRCh38
rs753830870 Chr.1: 6825180-6825180 on Build GRCh38
rs374514641 Chr.1: 6825086-6825086 on Build GRCh38
rs145982830 Chr.1: 6825138-6825138 on Build GRCh38
rs989147228 Chr.1: 6825132-6825132 on Build GRCh38
rs971952062 Chr.1: 6825084-6825084 on Build GRCh38
rs201916535 Chr.1: 6825162-6825162 on Build GRCh38
rs755971110 Chr.1: 6825053-6825053 on Build GRCh38
rs778073709 Chr.1: 6825095-6825095 on Build GRCh38
rs1024504108 Chr.1: 6825021-6825021 on Build GRCh38
rs758782676 Chr.1: 6825054-6825056 on Build GRCh38
rs773938997 Chr.1: 6825161-6825161 on Build GRCh38
rs766952396 Chr.1: 6825050-6825050 on Build GRCh38
rs781163086 Chr.1: 6825111-6825111 on Build GRCh38

View Full Product Details