Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6640120-6640357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3464421 c.1317G>A p.M439I Chr.12: 6640168-6640168 on Build GRCh38
4999878 c.1309C>A p.L437I Chr.12: 6640176-6640176 on Build GRCh38
4044368 c.1280G>A p.R427H Chr.12: 6640205-6640205 on Build GRCh38
2098758 c.1344G>A p.T448T Chr.12: 6640141-6640141 on Build GRCh38
6073369 c.1302C>T p.Y434Y Chr.12: 6640183-6640183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143853195 Chr.12: 6640198-6640198 on Build GRCh38
rs771778088 Chr.12: 6640323-6640323 on Build GRCh38
rs777709026 Chr.12: 6640182-6640182 on Build GRCh38
rs773425168 Chr.12: 6640207-6640207 on Build GRCh38
rs756182832 Chr.12: 6640170-6640170 on Build GRCh38
rs749150311 Chr.12: 6640183-6640183 on Build GRCh38
rs371102886 Chr.12: 6640152-6640152 on Build GRCh38
rs114920773 Chr.12: 6640205-6640205 on Build GRCh38
rs768133121 Chr.12: 6640234-6640234 on Build GRCh38
rs572932109 Chr.12: 6640210-6640210 on Build GRCh38
rs1048374371 Chr.12: 6640315-6640315 on Build GRCh38
rs770439662 Chr.12: 6640195-6640195 on Build GRCh38
rs373845141 Chr.12: 6640274-6640274 on Build GRCh38
rs758246686 Chr.12: 6640165-6640165 on Build GRCh38
rs905253039 Chr.12: 6640263-6640263 on Build GRCh38
rs752895446 Chr.12: 6640243-6640243 on Build GRCh38
rs371593728 Chr.12: 6640332-6640332 on Build GRCh38
rs201606879 Chr.12: 6640331-6640331 on Build GRCh38
rs776336522 Chr.12: 6640271-6640273 on Build GRCh38
rs150050921 Chr.12: 6640222-6640222 on Build GRCh38
rs375599035 Chr.12: 6640234-6640234 on Build GRCh38
rs766471960 Chr.12: 6640218-6640218 on Build GRCh38
rs756092794 Chr.12: 6640246-6640246 on Build GRCh38
rs71450161 Chr.12: 6640271-6640271 on Build GRCh38
rs781209644 Chr.12: 6640143-6640143 on Build GRCh38
rs368274050 Chr.12: 6640256-6640256 on Build GRCh38
rs913740855 Chr.12: 6640253-6640253 on Build GRCh38
rs372480201 Chr.12: 6640260-6640260 on Build GRCh38
rs146172116 Chr.12: 6640141-6640141 on Build GRCh38
rs748992833 Chr.12: 6640257-6640257 on Build GRCh38
rs745505796 Chr.12: 6640321-6640321 on Build GRCh38
rs140185383 Chr.12: 6640197-6640197 on Build GRCh38
rs767695307 Chr.12: 6640241-6640241 on Build GRCh38
rs543415532 Chr.12: 6640287-6640287 on Build GRCh38
rs553167752 Chr.12: 6640206-6640206 on Build GRCh38
rs922988124 Chr.12: 6640330-6640330 on Build GRCh38

View Full Product Details