Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84195530-84195785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAD2
Entrez Gene ID
161931
Gene Name
adenosine deaminase domain containing 2
Gene Aliases
TENRL
UniGene
Hs.8977
Gene Chromosome Location
Chr.16: 84191117-84197166 on Build GRCh38
Transcripts
NM_001145400.1 NM_139174.3 XM_005255814.1 XM_011522919.1 XM_011522920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6593323 c.1192G>A p.V398M Chr.16: 84195591-84195591 on Build GRCh38
974369 c.1289G>A p.R430H Chr.16: 84195688-84195688 on Build GRCh38
6593322 c.1145A>G p.Q382R Chr.16: 84195544-84195544 on Build GRCh38
7202030 c.1192G>T p.V398L Chr.16: 84195591-84195591 on Build GRCh38
472184 c.1243G>T p.V415F Chr.16: 84195642-84195642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763857671 Chr.16: 84195608-84195608 on Build GRCh38
rs758700262 Chr.16: 84195707-84195708 on Build GRCh38
rs755276888 Chr.16: 84195643-84195643 on Build GRCh38
rs367895841 Chr.16: 84195683-84195683 on Build GRCh38
rs759342116 Chr.16: 84195632-84195632 on Build GRCh38
rs760330356 Chr.16: 84195549-84195549 on Build GRCh38
rs769869574 Chr.16: 84195635-84195635 on Build GRCh38
rs200468657 Chr.16: 84195626-84195626 on Build GRCh38
rs780330737 Chr.16: 84195613-84195613 on Build GRCh38
rs751608071 Chr.16: 84195725-84195725 on Build GRCh38
rs962393185 Chr.16: 84195692-84195692 on Build GRCh38
rs780639899 Chr.16: 84195567-84195567 on Build GRCh38
rs778291994 Chr.16: 84195571-84195571 on Build GRCh38
rs759453180 Chr.16: 84195595-84195595 on Build GRCh38
rs139952223 Chr.16: 84195682-84195682 on Build GRCh38
rs371012828 Chr.16: 84195640-84195640 on Build GRCh38
rs547168217 Chr.16: 84195681-84195681 on Build GRCh38
rs923799301 Chr.16: 84195562-84195562 on Build GRCh38
rs141930279 Chr.16: 84195629-84195629 on Build GRCh38
rs763031674 Chr.16: 84195688-84195688 on Build GRCh38
rs768622169 Chr.16: 84195698-84195698 on Build GRCh38
rs772099322 Chr.16: 84195617-84195617 on Build GRCh38
rs758881760 Chr.16: 84195665-84195665 on Build GRCh38
rs1036541885 Chr.16: 84195753-84195753 on Build GRCh38
rs202063579 Chr.16: 84195638-84195638 on Build GRCh38
rs372591388 Chr.16: 84195740-84195740 on Build GRCh38
rs201627932 Chr.16: 84195654-84195654 on Build GRCh38
rs772285105 Chr.16: 84195582-84195582 on Build GRCh38
rs760289874 Chr.16: 84195708-84195708 on Build GRCh38
rs758193797 Chr.16: 84195562-84195562 on Build GRCh38
rs766363314 Chr.16: 84195551-84195551 on Build GRCh38
rs763550117 Chr.16: 84195639-84195639 on Build GRCh38
rs201035894 Chr.16: 84195615-84195615 on Build GRCh38
rs765641028 Chr.16: 84195712-84195712 on Build GRCh38
rs955307081 Chr.16: 84195563-84195563 on Build GRCh38
rs920330552 Chr.16: 84195755-84195755 on Build GRCh38
rs576824503 Chr.16: 84195731-84195731 on Build GRCh38
rs768481485 Chr.16: 84195741-84195741 on Build GRCh38
rs777880070 Chr.16: 84195618-84195618 on Build GRCh38
rs749796902 Chr.16: 84195568-84195568 on Build GRCh38
rs773259201 Chr.16: 84195584-84195584 on Build GRCh38
rs191155110 Chr.16: 84195703-84195703 on Build GRCh38
rs746531516 Chr.16: 84195614-84195614 on Build GRCh38
rs756899996 Chr.16: 84195612-84195612 on Build GRCh38
rs376317175 Chr.16: 84195717-84195717 on Build GRCh38
rs753242775 Chr.16: 84195659-84195659 on Build GRCh38
rs1056004499 Chr.16: 84195764-84195764 on Build GRCh38
rs146308441 Chr.16: 84195680-84195680 on Build GRCh38
rs752245764 Chr.16: 84195561-84195561 on Build GRCh38
rs764892126 Chr.16: 84195560-84195560 on Build GRCh38
rs377399416 Chr.16: 84195594-84195594 on Build GRCh38
rs116988957 Chr.16: 84195762-84195762 on Build GRCh38
rs200245527 Chr.16: 84195700-84195700 on Build GRCh38
rs762498754 Chr.16: 84195602-84195602 on Build GRCh38
rs751128927 Chr.16: 84195609-84195609 on Build GRCh38
rs562445876 Chr.16: 84195622-84195622 on Build GRCh38
rs754913696 Chr.16: 84195557-84195557 on Build GRCh38
rs950151982 Chr.16: 84195746-84195746 on Build GRCh38
rs764597926 Chr.16: 84195722-84195722 on Build GRCh38
rs747111471 Chr.16: 84195675-84195675 on Build GRCh38
rs774469734 Chr.16: 84195569-84195569 on Build GRCh38
rs746128962 Chr.16: 84195736-84195736 on Build GRCh38
rs1042220636 Chr.16: 84195747-84195747 on Build GRCh38
rs150649614 Chr.16: 84195566-84195566 on Build GRCh38
rs143488002 Chr.16: 84195687-84195687 on Build GRCh38
rs541146214 Chr.16: 84195586-84195586 on Build GRCh38
rs368334407 Chr.16: 84195721-84195721 on Build GRCh38
rs903222318 Chr.16: 84195636-84195636 on Build GRCh38
rs920812984 Chr.16: 84195710-84195710 on Build GRCh38
rs757315128 Chr.16: 84195726-84195726 on Build GRCh38
rs754158131 Chr.16: 84195642-84195642 on Build GRCh38
rs756379335 Chr.16: 84195737-84195737 on Build GRCh38
rs767089179 Chr.16: 84195702-84195702 on Build GRCh38
rs772470445 Chr.16: 84195564-84195564 on Build GRCh38
rs748221639 Chr.16: 84195580-84195580 on Build GRCh38
rs761429242 Chr.16: 84195641-84195641 on Build GRCh38
rs750742878 Chr.16: 84195562-84195563 on Build GRCh38
rs138702097 Chr.16: 84195601-84195601 on Build GRCh38
rs1036102053 Chr.16: 84195606-84195606 on Build GRCh38
rs766129197 Chr.16: 84195591-84195591 on Build GRCh38
rs765263888 Chr.16: 84195596-84195596 on Build GRCh38
rs753048433 Chr.16: 84195716-84195716 on Build GRCh38
rs753678549 Chr.16: 84195554-84195554 on Build GRCh38

View Full Product Details