Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 10461280-10461553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD3
Entrez Gene ID
272
Gene Name
adenosine monophosphate deaminase 3
Gene Aliases
-
UniGene
Hs.501890
Gene Chromosome Location
Chr.11: 10450321-10507579 on Build GRCh38
Transcripts
NM_000480.2 NM_001025389.1 NM_001025390.1 NM_001172430.1 NM_001172431.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297582 c.31C>T p.P11S Chr.11: 10461523-10461523 on Build GRCh38
6599439 c.32C>T p.P11L Chr.11: 10461524-10461524 on Build GRCh38
5422178 c.34C>T p.R12W Chr.11: 10461526-10461526 on Build GRCh38
1675840 c.35G>A p.R12Q Chr.11: 10461527-10461527 on Build GRCh38
3791120 c.51G>A p.L17L Chr.11: 10461543-10461543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751921390 Chr.11: 10461446-10461446 on Build GRCh38
rs201661559 Chr.11: 10461437-10461437 on Build GRCh38
rs767955616 Chr.11: 10461458-10461458 on Build GRCh38
rs540683532 Chr.11: 10461509-10461509 on Build GRCh38
rs115759384 Chr.11: 10461374-10461374 on Build GRCh38
rs372846179 Chr.11: 10461424-10461424 on Build GRCh38
rs145604316 Chr.11: 10461524-10461524 on Build GRCh38
rs576749734 Chr.11: 10461445-10461445 on Build GRCh38
rs776533422 Chr.11: 10461408-10461408 on Build GRCh38
rs745366158 Chr.11: 10461481-10461481 on Build GRCh38
rs773226493 Chr.11: 10461515-10461515 on Build GRCh38
rs746879134 Chr.11: 10461489-10461489 on Build GRCh38
rs78155698 Chr.11: 10461461-10461461 on Build GRCh38
rs763501693 Chr.11: 10461407-10461407 on Build GRCh38
rs543879425 Chr.11: 10461323-10461324 on Build GRCh38
rs867446911 Chr.11: 10461523-10461523 on Build GRCh38
rs778629569 Chr.11: 10461462-10461462 on Build GRCh38
rs561207052 Chr.11: 10461525-10461525 on Build GRCh38
rs775206482 Chr.11: 10461416-10461416 on Build GRCh38
rs545122288 Chr.11: 10461447-10461447 on Build GRCh38
rs372097330 Chr.11: 10461487-10461487 on Build GRCh38
rs978280311 Chr.11: 10461382-10461382 on Build GRCh38
rs910036380 Chr.11: 10461315-10461315 on Build GRCh38
rs763070134 Chr.11: 10461519-10461519 on Build GRCh38
rs149625321 Chr.11: 10461428-10461428 on Build GRCh38
rs984392653 Chr.11: 10461308-10461308 on Build GRCh38
rs1036778591 Chr.11: 10461326-10461326 on Build GRCh38
rs778505193 Chr.11: 10461412-10461412 on Build GRCh38
rs186250080 Chr.11: 10461429-10461429 on Build GRCh38
rs148904594 Chr.11: 10461526-10461526 on Build GRCh38
rs527260375 Chr.11: 10461503-10461503 on Build GRCh38
rs147321717 Chr.11: 10461434-10461434 on Build GRCh38
rs568339118 Chr.11: 10461316-10461316 on Build GRCh38
rs753551502 Chr.11: 10461460-10461460 on Build GRCh38
rs376360074 Chr.11: 10461505-10461505 on Build GRCh38
rs933741878 Chr.11: 10461339-10461339 on Build GRCh38
rs761179959 Chr.11: 10461486-10461486 on Build GRCh38
rs745794355 Chr.11: 10461414-10461414 on Build GRCh38
rs1049363661 Chr.11: 10461371-10461371 on Build GRCh38
rs138271082 Chr.11: 10461406-10461406 on Build GRCh38
rs768905591 Chr.11: 10461427-10461427 on Build GRCh38
rs763939370 Chr.11: 10461379-10461379 on Build GRCh38
rs945144462 Chr.11: 10461377-10461377 on Build GRCh38
rs557880170 Chr.11: 10461338-10461338 on Build GRCh38

View Full Product Details