Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 55158335-55158592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP5
Entrez Gene ID
9787
Gene Name
DLG associated protein 5
Gene Aliases
DLG7, HURP
UniGene
Hs.77695
Gene Chromosome Location
Chr.14: 55148116-55191678 on Build GRCh38
Transcripts
NM_001146015.1 NM_014750.4 XM_017021840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
171990 c.1827C>T p.F609F Chr.14: 55158568-55158568 on Build GRCh38
5624269 c.1856C>G p.P619R Chr.14: 55158539-55158539 on Build GRCh38
4975301 c.1842C>T p.F614F Chr.14: 55158553-55158553 on Build GRCh38
470036 c.1803A>G p.P601P Chr.14: 55158592-55158592 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545178100 Chr.14: 55158406-55158406 on Build GRCh38
rs746686691 Chr.14: 55158500-55158500 on Build GRCh38
rs778125741 Chr.14: 55158477-55158477 on Build GRCh38
rs1050558464 Chr.14: 55158367-55158367 on Build GRCh38
rs746040103 Chr.14: 55158500-55158500 on Build GRCh38
rs546157515 Chr.14: 55158501-55158501 on Build GRCh38
rs993836067 Chr.14: 55158450-55158450 on Build GRCh38
rs200878291 Chr.14: 55158508-55158508 on Build GRCh38
rs766073374 Chr.14: 55158512-55158514 on Build GRCh38
rs542092291 Chr.14: 55158452-55158452 on Build GRCh38
rs145664806 Chr.14: 55158510-55158510 on Build GRCh38
rs749868690 Chr.14: 55158478-55158478 on Build GRCh38
rs912160514 Chr.14: 55158368-55158368 on Build GRCh38
rs888174777 Chr.14: 55158457-55158457 on Build GRCh38
rs774007246 Chr.14: 55158542-55158542 on Build GRCh38
rs778975358 Chr.14: 55158496-55158496 on Build GRCh38
rs12881211 Chr.14: 55158409-55158409 on Build GRCh38
rs1018720255 Chr.14: 55158442-55158442 on Build GRCh38
rs1036614701 Chr.14: 55158389-55158389 on Build GRCh38
rs1012049689 Chr.14: 55158466-55158466 on Build GRCh38
rs1006866071 Chr.14: 55158404-55158405 on Build GRCh38
rs952791015 Chr.14: 55158402-55158402 on Build GRCh38
rs372360305 Chr.14: 55158511-55158511 on Build GRCh38
rs12882439 Chr.14: 55158408-55158408 on Build GRCh38
rs139156235 Chr.14: 55158491-55158492 on Build GRCh38
rs57075984 Chr.14: 55158417-55158417 on Build GRCh38
rs768242490 Chr.14: 55158515-55158518 on Build GRCh38
rs553139401 Chr.14: 55158375-55158375 on Build GRCh38
rs761825301 Chr.14: 55158509-55158509 on Build GRCh38
rs368248560 Chr.14: 55158486-55158486 on Build GRCh38
rs896847919 Chr.14: 55158445-55158445 on Build GRCh38
rs111324853 Chr.14: 55158475-55158475 on Build GRCh38

View Full Product Details