Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99840146-99840417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPP1
Entrez Gene ID
27071
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
Gene Aliases
BAM32
UniGene
Hs.436271
Gene Chromosome Location
Chr.4: 99816824-99872290 on Build GRCh38
Transcripts
NM_001306151.1 NM_014395.2 XM_011531840.2 XM_011531842.1 XM_017008023.1 XM_017008024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4438969 c.321G>T p.K107N Chr.4: 99840385-99840385 on Build GRCh38
7245728 c.225-1G>T p.? Chr.4: 99840288-99840288 on Build GRCh38
5759132 c.342G>C p.L114F Chr.4: 99840406-99840406 on Build GRCh38
5653255 c.272C>A p.S91* Chr.4: 99840336-99840336 on Build GRCh38
1049455 c.263C>T p.T88I Chr.4: 99840327-99840327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761021574 Chr.4: 99840249-99840249 on Build GRCh38
rs74586862 Chr.4: 99840225-99840225 on Build GRCh38
rs763855611 Chr.4: 99840358-99840358 on Build GRCh38
rs541741416 Chr.4: 99840315-99840315 on Build GRCh38
rs1012162011 Chr.4: 99840190-99840190 on Build GRCh38
rs967467199 Chr.4: 99840215-99840215 on Build GRCh38
rs531841086 Chr.4: 99840290-99840290 on Build GRCh38
rs758320947 Chr.4: 99840286-99840286 on Build GRCh38
rs773506907 Chr.4: 99840242-99840242 on Build GRCh38
rs766591104 Chr.4: 99840263-99840263 on Build GRCh38
rs1021629471 Chr.4: 99840207-99840207 on Build GRCh38
rs77394557 Chr.4: 99840219-99840219 on Build GRCh38
rs567264365 Chr.4: 99840253-99840253 on Build GRCh38
rs377247197 Chr.4: 99840282-99840282 on Build GRCh38
rs552764635 Chr.4: 99840271-99840271 on Build GRCh38
rs564612149 Chr.4: 99840275-99840275 on Build GRCh38
rs559809943 Chr.4: 99840368-99840368 on Build GRCh38
rs937899470 Chr.4: 99840336-99840336 on Build GRCh38
rs781011074 Chr.4: 99840355-99840355 on Build GRCh38
rs757057345 Chr.4: 99840332-99840332 on Build GRCh38
rs977387118 Chr.4: 99840217-99840217 on Build GRCh38
rs267599964 Chr.4: 99840312-99840312 on Build GRCh38
rs752794502 Chr.4: 99840283-99840283 on Build GRCh38
rs976350867 Chr.4: 99840173-99840173 on Build GRCh38

View Full Product Details