Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143611354-143611581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM3
Entrez Gene ID
166752
Gene Name
FRAS1 related extracellular matrix 3
Gene Aliases
-
UniGene
Hs.252714
Gene Chromosome Location
Chr.4: 143577407-143700675 on Build GRCh38
Transcripts
NM_001168235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5636146 c.5835T>C p.R1945R Chr.4: 143611472-143611472 on Build GRCh38
5060145 c.5799T>C p.I1933I Chr.4: 143611508-143611508 on Build GRCh38
5644344 c.5802T>A p.S1934S Chr.4: 143611505-143611505 on Build GRCh38
6199624 c.5891G>T p.C1964F Chr.4: 143611416-143611416 on Build GRCh38
5692107 c.5844C>A p.D1948E Chr.4: 143611463-143611463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866299750 Chr.4: 143611486-143611486 on Build GRCh38
rs545010807 Chr.4: 143611415-143611415 on Build GRCh38
rs768167367 Chr.4: 143611527-143611527 on Build GRCh38
rs72938299 Chr.4: 143611491-143611491 on Build GRCh38
rs755221141 Chr.4: 143611378-143611378 on Build GRCh38
rs72938301 Chr.4: 143611540-143611540 on Build GRCh38
rs781351513 Chr.4: 143611381-143611381 on Build GRCh38
rs368852203 Chr.4: 143611384-143611384 on Build GRCh38
rs572037223 Chr.4: 143611473-143611473 on Build GRCh38
rs990782846 Chr.4: 143611522-143611522 on Build GRCh38
rs961893791 Chr.4: 143611416-143611416 on Build GRCh38
rs560006254 Chr.4: 143611438-143611438 on Build GRCh38
rs200203147 Chr.4: 143611474-143611474 on Build GRCh38
rs766506285 Chr.4: 143611400-143611400 on Build GRCh38
rs145310584 Chr.4: 143611543-143611543 on Build GRCh38
rs577569170 Chr.4: 143611409-143611409 on Build GRCh38
rs770110045 Chr.4: 143611546-143611546 on Build GRCh38
rs757082007 Chr.4: 143611403-143611403 on Build GRCh38
rs1016176307 Chr.4: 143611404-143611404 on Build GRCh38
rs961124792 Chr.4: 143611426-143611426 on Build GRCh38
rs752993629 Chr.4: 143611392-143611392 on Build GRCh38

View Full Product Details