Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67419482-67419733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARNS1
Entrez Gene ID
57571
Gene Name
carnosine synthase 1
Gene Aliases
ATPGD1
UniGene
Hs.502982
Gene Chromosome Location
Chr.11: 67415678-67425607 on Build GRCh38
Transcripts
NM_001166222.1 NM_020811.1 XM_011545189.2 XM_011545191.2 XM_017018054.1 XM_017018055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3869990 c.904C>T p.L302L Chr.11: 67419538-67419538 on Build GRCh38
4647425 c.997G>A p.V333M Chr.11: 67419631-67419631 on Build GRCh38
1356397 c.163C>A p.Q55K Chr.11: 67419591-67419591 on Build GRCh38
6495981 c.1007C>T p.P336L Chr.11: 67419641-67419641 on Build GRCh38
5839372 c.991G>A p.E331K Chr.11: 67419625-67419625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778084627 Chr.11: 67419694-67419694 on Build GRCh38
rs767706133 Chr.11: 67419596-67419604 on Build GRCh38
rs772413089 Chr.11: 67419564-67419564 on Build GRCh38
rs371047026 Chr.11: 67419584-67419584 on Build GRCh38
rs375626867 Chr.11: 67419547-67419547 on Build GRCh38
rs749185188 Chr.11: 67419681-67419682 on Build GRCh38
rs758285713 Chr.11: 67419543-67419543 on Build GRCh38
rs199710256 Chr.11: 67419703-67419703 on Build GRCh38
rs41302427 Chr.11: 67419524-67419524 on Build GRCh38
rs1055992384 Chr.11: 67419534-67419534 on Build GRCh38
rs1007786136 Chr.11: 67419562-67419562 on Build GRCh38
rs757410786 Chr.11: 67419515-67419520 on Build GRCh38
rs752383746 Chr.11: 67419599-67419599 on Build GRCh38
rs370587703 Chr.11: 67419518-67419518 on Build GRCh38
rs370411760 Chr.11: 67419540-67419540 on Build GRCh38
rs746941221 Chr.11: 67419545-67419545 on Build GRCh38
rs887962861 Chr.11: 67419535-67419535 on Build GRCh38
rs756131332 Chr.11: 67419640-67419652 on Build GRCh38
rs568921519 Chr.11: 67419548-67419548 on Build GRCh38
rs773632607 Chr.11: 67419565-67419565 on Build GRCh38
rs764741356 Chr.11: 67419592-67419592 on Build GRCh38
rs781222928 Chr.11: 67419653-67419653 on Build GRCh38
rs375777771 Chr.11: 67419644-67419644 on Build GRCh38
rs755096802 Chr.11: 67419662-67419662 on Build GRCh38
rs1021683934 Chr.11: 67419560-67419560 on Build GRCh38
rs374499029 Chr.11: 67419546-67419546 on Build GRCh38
rs958124077 Chr.11: 67419593-67419593 on Build GRCh38
rs774402993 Chr.11: 67419600-67419602 on Build GRCh38
rs1046139654 Chr.11: 67419512-67419512 on Build GRCh38
rs761136107 Chr.11: 67419566-67419566 on Build GRCh38
rs1022818732 Chr.11: 67419609-67419609 on Build GRCh38
rs778916585 Chr.11: 67419671-67419671 on Build GRCh38
rs370181519 Chr.11: 67419640-67419640 on Build GRCh38
rs767944697 Chr.11: 67419511-67419511 on Build GRCh38
rs367988733 Chr.11: 67419674-67419674 on Build GRCh38
rs763953799 Chr.11: 67419618-67419618 on Build GRCh38
rs937496663 Chr.11: 67419530-67419530 on Build GRCh38
rs765868422 Chr.11: 67419523-67419523 on Build GRCh38
rs376841520 Chr.11: 67419704-67419704 on Build GRCh38
rs570844079 Chr.11: 67419517-67419517 on Build GRCh38
rs747395059 Chr.11: 67419695-67419695 on Build GRCh38
rs375601665 Chr.11: 67419536-67419536 on Build GRCh38
rs368884705 Chr.11: 67419585-67419585 on Build GRCh38
rs768973886 Chr.11: 67419503-67419503 on Build GRCh38
rs79669706 Chr.11: 67419698-67419698 on Build GRCh38
rs772319838 Chr.11: 67419675-67419675 on Build GRCh38
rs762270521 Chr.11: 67419506-67419506 on Build GRCh38
rs771944922 Chr.11: 67419648-67419655 on Build GRCh38
rs370760192 Chr.11: 67419601-67419601 on Build GRCh38
rs749260663 Chr.11: 67419657-67419657 on Build GRCh38

View Full Product Details