Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15380316-15380557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
UniGene
Hs.399800
Gene Chromosome Location
Chr.19: 15380048-15419121 on Build GRCh38
Transcripts
NM_001291478.1 NM_014371.3 NR_111971.1 XM_005259854.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438778 c.1625A>G p.Y542C Chr.19: 15380524-15380524 on Build GRCh38
1391108 c.1666_1668delGAG p.E556delE Chr.19: 15380395-15380397 on Build GRCh38
5854854 c.1618G>A p.E540K Chr.19: 15380531-15380531 on Build GRCh38
6375942 c.1744G>A p.A582T Chr.19: 15380319-15380319 on Build GRCh38
5668502 c.1688A>T p.E563V Chr.19: 15380375-15380375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372789644 Chr.19: 15380437-15380437 on Build GRCh38
rs368820903 Chr.19: 15380386-15380386 on Build GRCh38
rs1041427583 Chr.19: 15380388-15380389 on Build GRCh38
rs762780905 Chr.19: 15380427-15380427 on Build GRCh38
rs989194293 Chr.19: 15380512-15380512 on Build GRCh38
rs770530550 Chr.19: 15380412-15380412 on Build GRCh38
rs763186493 Chr.19: 15380368-15380368 on Build GRCh38
rs749073857 Chr.19: 15380502-15380502 on Build GRCh38
rs751611212 Chr.19: 15380377-15380377 on Build GRCh38
rs764244285 Chr.19: 15380472-15380473 on Build GRCh38
rs745833631 Chr.19: 15380421-15380421 on Build GRCh38
rs762728855 Chr.19: 15380459-15380460 on Build GRCh38
rs780778809 Chr.19: 15380471-15380471 on Build GRCh38
rs755443343 Chr.19: 15380454-15380454 on Build GRCh38
rs1055596462 Chr.19: 15380470-15380470 on Build GRCh38
rs777608722 Chr.19: 15380404-15380404 on Build GRCh38
rs745753337 Chr.19: 15380494-15380494 on Build GRCh38
rs530771954 Chr.19: 15380373-15380373 on Build GRCh38
rs746495361 Chr.19: 15380406-15380406 on Build GRCh38
rs751008965 Chr.19: 15380383-15380383 on Build GRCh38
rs763823019 Chr.19: 15380458-15380458 on Build GRCh38
rs775490570 Chr.19: 15380395-15380397 on Build GRCh38
rs767901026 Chr.19: 15380381-15380381 on Build GRCh38
rs780925811 Chr.19: 15380415-15380415 on Build GRCh38
rs750836113 Chr.19: 15380440-15380440 on Build GRCh38
rs765234638 Chr.19: 15380333-15380333 on Build GRCh38
rs748413083 Chr.19: 15380528-15380528 on Build GRCh38
rs756014939 Chr.19: 15380497-15380497 on Build GRCh38
rs911387823 Chr.19: 15380363-15380363 on Build GRCh38
rs751291976 Chr.19: 15380465-15380465 on Build GRCh38
rs769427887 Chr.19: 15380424-15380424 on Build GRCh38
rs145945826 Chr.19: 15380497-15380498 on Build GRCh38
rs761263248 Chr.19: 15380442-15380442 on Build GRCh38
rs767636501 Chr.19: 15380439-15380439 on Build GRCh38
rs756665695 Chr.19: 15380468-15380468 on Build GRCh38
rs374438016 Chr.19: 15380508-15380508 on Build GRCh38
rs776678564 Chr.19: 15380332-15380332 on Build GRCh38
rs769638010 Chr.19: 15380364-15380364 on Build GRCh38
rs774616750 Chr.19: 15380435-15380435 on Build GRCh38
rs202121058 Chr.19: 15380398-15380398 on Build GRCh38
rs752492853 Chr.19: 15380387-15380387 on Build GRCh38
rs768848516 Chr.19: 15380428-15380428 on Build GRCh38
rs771993469 Chr.19: 15380529-15380529 on Build GRCh38
rs775295991 Chr.19: 15380425-15380425 on Build GRCh38
rs1047694815 Chr.19: 15380346-15380346 on Build GRCh38
rs775633249 Chr.19: 15380367-15380367 on Build GRCh38
rs759236696 Chr.19: 15380340-15380340 on Build GRCh38
rs762155709 Chr.19: 15380496-15380497 on Build GRCh38
rs774492314 Chr.19: 15380523-15380523 on Build GRCh38
rs766806937 Chr.19: 15380452-15380452 on Build GRCh38
rs1000207351 Chr.19: 15380443-15380443 on Build GRCh38
rs544609212 Chr.19: 15380498-15380498 on Build GRCh38
rs774524128 Chr.19: 15380484-15380485 on Build GRCh38

View Full Product Details