Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47679269-47679522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
UniGene
Hs.97244
Gene Chromosome Location
Chr.19: 47608196-47703277 on Build GRCh38
Transcripts
NM_015711.3 XM_005258833.4 XM_006723180.3 XM_011526882.2 XM_011526883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6364833 c.160C>A p.Q54K Chr.19: 47679330-47679330 on Build GRCh38
7369583 c.202C>T p.P68S Chr.19: 47679372-47679372 on Build GRCh38
1394928 c.258G>A p.A86A Chr.19: 47679428-47679428 on Build GRCh38
5968120 c.278G>T p.G93V Chr.19: 47679448-47679448 on Build GRCh38
4543765 c.343G>A p.E115K Chr.19: 47679513-47679513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776467906 Chr.19: 47679325-47679325 on Build GRCh38
rs886736722 Chr.19: 47679458-47679458 on Build GRCh38
rs191148484 Chr.19: 47679441-47679441 on Build GRCh38
rs911200431 Chr.19: 47679459-47679459 on Build GRCh38
rs889219529 Chr.19: 47679434-47679434 on Build GRCh38
rs2911005 Chr.19: 47679311-47679311 on Build GRCh38
rs752723434 Chr.19: 47679433-47679433 on Build GRCh38
rs942604977 Chr.19: 47679460-47679460 on Build GRCh38
rs138544372 Chr.19: 47679440-47679440 on Build GRCh38
rs1052849514 Chr.19: 47679347-47679347 on Build GRCh38
rs909639054 Chr.19: 47679298-47679298 on Build GRCh38
rs757546071 Chr.19: 47679443-47679443 on Build GRCh38
rs762884075 Chr.19: 47679395-47679395 on Build GRCh38
rs768403190 Chr.19: 47679315-47679315 on Build GRCh38
rs766259836 Chr.19: 47679427-47679427 on Build GRCh38
rs199776123 Chr.19: 47679382-47679382 on Build GRCh38
rs913936553 Chr.19: 47679351-47679351 on Build GRCh38
rs893513555 Chr.19: 47679300-47679300 on Build GRCh38
rs765138718 Chr.19: 47679341-47679341 on Build GRCh38
rs554531895 Chr.19: 47679290-47679290 on Build GRCh38
rs147126463 Chr.19: 47679332-47679332 on Build GRCh38
rs779275819 Chr.19: 47679444-47679444 on Build GRCh38
rs772895921 Chr.19: 47679374-47679374 on Build GRCh38
rs1005184849 Chr.19: 47679470-47679470 on Build GRCh38
rs1052597969 Chr.19: 47679294-47679294 on Build GRCh38
rs946779321 Chr.19: 47679420-47679420 on Build GRCh38
rs943510885 Chr.19: 47679437-47679437 on Build GRCh38
rs922454643 Chr.19: 47679504-47679504 on Build GRCh38
rs186397251 Chr.19: 47679310-47679310 on Build GRCh38
rs976650091 Chr.19: 47679467-47679467 on Build GRCh38
rs980642820 Chr.19: 47679318-47679318 on Build GRCh38
rs1035833505 Chr.19: 47679414-47679414 on Build GRCh38
rs556580334 Chr.19: 47679438-47679438 on Build GRCh38

View Full Product Details