Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45408785-45409032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3EAP
Entrez Gene ID
10849
Gene Name
CD3e molecule associated protein
Gene Aliases
ASE-1, ASE1, CAST, PAF49
UniGene
Hs.710495
Gene Chromosome Location
Chr.19: 45406129-45410766 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1 NM_001166049.1 NM_001983.3 XM_017026460.1 XM_017026461.1 XM_017026462.1 XM_017026463.1 XM_017026464.1 XM_017026465.1 XM_017026466.1

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
UniGene
Hs.435981
Gene Chromosome Location
Chr.19: 45407333-45478866 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1 NM_001166049.1 NM_001983.3 XM_017026460.1 XM_017026461.1 XM_017026462.1 XM_017026463.1 XM_017026464.1 XM_017026465.1 XM_017026466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998196 c.956A>G p.E319G Chr.19: 45408924-45408924 on Build GRCh38
82057 c.954G>A p.L318L Chr.19: 45408922-45408922 on Build GRCh38
6880519 c.852G>A p.L284L Chr.19: 45408820-45408820 on Build GRCh38
4518908 c.860_861CC>TT p.T287I Chr.19: 45408828-45408829 on Build GRCh38
1394606 c.1031C>T p.T344M Chr.19: 45408999-45408999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3212989 Chr.19: 45408812-45408812 on Build GRCh38
rs964222383 Chr.19: 45408948-45408948 on Build GRCh38
rs777247662 Chr.19: 45408823-45408823 on Build GRCh38
rs781466069 Chr.19: 45409001-45409001 on Build GRCh38
rs372074228 Chr.19: 45408827-45408827 on Build GRCh38
rs749947084 Chr.19: 45408987-45408987 on Build GRCh38
rs761911449 Chr.19: 45408869-45408869 on Build GRCh38
rs759389149 Chr.19: 45408832-45408837 on Build GRCh38
rs746379344 Chr.19: 45408913-45408913 on Build GRCh38
rs139303076 Chr.19: 45408826-45408826 on Build GRCh38
rs757999734 Chr.19: 45408991-45408991 on Build GRCh38
rs772550576 Chr.19: 45408852-45408852 on Build GRCh38
rs769960893 Chr.19: 45409006-45409006 on Build GRCh38
rs143986145 Chr.19: 45408855-45408855 on Build GRCh38
rs777368519 Chr.19: 45408901-45408901 on Build GRCh38
rs199895540 Chr.19: 45409000-45409000 on Build GRCh38
rs747329205 Chr.19: 45408928-45408928 on Build GRCh38
rs753518136 Chr.19: 45408960-45408960 on Build GRCh38
rs1018947921 Chr.19: 45408947-45408947 on Build GRCh38
rs371868030 Chr.19: 45408955-45408955 on Build GRCh38
rs199589536 Chr.19: 45408856-45408856 on Build GRCh38
rs760400069 Chr.19: 45408954-45408954 on Build GRCh38
rs751453778 Chr.19: 45408875-45408875 on Build GRCh38
rs767394181 Chr.19: 45408949-45408949 on Build GRCh38
rs774856339 Chr.19: 45408841-45408841 on Build GRCh38
rs143443617 Chr.19: 45408923-45408923 on Build GRCh38
rs773505594 Chr.19: 45409010-45409010 on Build GRCh38
rs748783248 Chr.19: 45408825-45408825 on Build GRCh38
rs748372404 Chr.19: 45409005-45409005 on Build GRCh38
rs371563856 Chr.19: 45408944-45408944 on Build GRCh38
rs780667352 Chr.19: 45408993-45408993 on Build GRCh38
rs1047124023 Chr.19: 45408817-45408817 on Build GRCh38
rs148669652 Chr.19: 45408871-45408871 on Build GRCh38
rs778451525 Chr.19: 45408984-45408984 on Build GRCh38
rs761952784 Chr.19: 45408936-45408936 on Build GRCh38
rs755925154 Chr.19: 45408899-45408899 on Build GRCh38
rs767269782 Chr.19: 45408884-45408884 on Build GRCh38
rs146750944 Chr.19: 45408999-45408999 on Build GRCh38
rs767701108 Chr.19: 45408828-45408828 on Build GRCh38
rs758456293 Chr.19: 45409006-45409029 on Build GRCh38
rs756736472 Chr.19: 45408909-45408909 on Build GRCh38
rs764460395 Chr.19: 45408861-45408861 on Build GRCh38
rs748900769 Chr.19: 45408902-45408902 on Build GRCh38
rs766725989 Chr.19: 45408906-45408906 on Build GRCh38
rs778346767 Chr.19: 45408912-45408912 on Build GRCh38
rs752931566 Chr.19: 45408949-45408951 on Build GRCh38
rs773206067 Chr.19: 45408943-45408943 on Build GRCh38
rs866543758 Chr.19: 45408992-45408992 on Build GRCh38
rs895475604 Chr.19: 45408881-45408881 on Build GRCh38
rs754939987 Chr.19: 45408878-45408878 on Build GRCh38
rs752666450 Chr.19: 45408952-45408952 on Build GRCh38
rs776844788 Chr.19: 45408935-45408935 on Build GRCh38
rs200618543 Chr.19: 45408916-45408916 on Build GRCh38
rs769173238 Chr.19: 45408934-45408934 on Build GRCh38
rs776736875 Chr.19: 45408867-45408867 on Build GRCh38
rs3212988 Chr.19: 45408886-45408886 on Build GRCh38
rs769915095 Chr.19: 45408941-45408941 on Build GRCh38
rs746376502 Chr.19: 45408843-45408843 on Build GRCh38
rs756925223 Chr.19: 45408979-45408979 on Build GRCh38
rs765443575 Chr.19: 45408836-45408839 on Build GRCh38

View Full Product Details