Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65155627-65155900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPX
Entrez Gene ID
10845
Gene Name
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases
-
UniGene
Hs.113823
Gene Chromosome Location
Chr.15: 65148219-65185420 on Build GRCh38
Transcripts
NM_006660.4 NR_133680.1 XM_011521164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4893150 c.1186C>T p.P396S Chr.15: 65155817-65155817 on Build GRCh38
5500129 c.1181A>G p.N394S Chr.15: 65155822-65155822 on Build GRCh38
964008 c.1189G>A p.E397K Chr.15: 65155814-65155814 on Build GRCh38
7123371 c.1202G>A p.R401Q Chr.15: 65155801-65155801 on Build GRCh38
5849548 c.1209_1210CC>TT p.R404C Chr.15: 65155793-65155794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747996124 Chr.15: 65155801-65155801 on Build GRCh38
rs1008172172 Chr.15: 65155716-65155716 on Build GRCh38
rs373886925 Chr.15: 65155725-65155725 on Build GRCh38
rs750476540 Chr.15: 65155822-65155822 on Build GRCh38
rs762356000 Chr.15: 65155804-65155804 on Build GRCh38
rs765298278 Chr.15: 65155698-65155698 on Build GRCh38
rs772728938 Chr.15: 65155764-65155764 on Build GRCh38
rs765256546 Chr.15: 65155807-65155807 on Build GRCh38
rs1027160427 Chr.15: 65155774-65155774 on Build GRCh38
rs761104857 Chr.15: 65155773-65155773 on Build GRCh38
rs149761898 Chr.15: 65155775-65155775 on Build GRCh38
rs774216614 Chr.15: 65155655-65155658 on Build GRCh38
rs758590719 Chr.15: 65155842-65155842 on Build GRCh38
rs149716210 Chr.15: 65155659-65155659 on Build GRCh38
rs112493401 Chr.15: 65155765-65155765 on Build GRCh38
rs764044206 Chr.15: 65155670-65155670 on Build GRCh38
rs765559841 Chr.15: 65155853-65155854 on Build GRCh38
rs1018336093 Chr.15: 65155721-65155721 on Build GRCh38
rs1025088963 Chr.15: 65155675-65155675 on Build GRCh38
rs754032262 Chr.15: 65155688-65155688 on Build GRCh38
rs775880805 Chr.15: 65155671-65155671 on Build GRCh38
rs767792257 Chr.15: 65155668-65155668 on Build GRCh38
rs141108125 Chr.15: 65155707-65155707 on Build GRCh38
rs775764773 Chr.15: 65155772-65155772 on Build GRCh38
rs757400794 Chr.15: 65155849-65155850 on Build GRCh38
rs751422227 Chr.15: 65155861-65155861 on Build GRCh38
rs753977356 Chr.15: 65155708-65155710 on Build GRCh38
rs1010059618 Chr.15: 65155662-65155662 on Build GRCh38
rs772744441 Chr.15: 65155655-65155655 on Build GRCh38
rs758598477 Chr.15: 65155713-65155713 on Build GRCh38
rs759478390 Chr.15: 65155655-65155663 on Build GRCh38
rs370149464 Chr.15: 65155672-65155672 on Build GRCh38
rs754958321 Chr.15: 65155736-65155736 on Build GRCh38
rs764673178 Chr.15: 65155682-65155682 on Build GRCh38
rs769306600 Chr.15: 65155760-65155760 on Build GRCh38
rs748010872 Chr.15: 65155755-65155755 on Build GRCh38
rs777283892 Chr.15: 65155776-65155776 on Build GRCh38
rs781102222 Chr.15: 65155752-65155752 on Build GRCh38
rs770581618 Chr.15: 65155766-65155766 on Build GRCh38
rs146800403 Chr.15: 65155851-65155851 on Build GRCh38
rs144638772 Chr.15: 65155694-65155694 on Build GRCh38
rs147893822 Chr.15: 65155732-65155732 on Build GRCh38
rs762607840 Chr.15: 65155660-65155660 on Build GRCh38
rs952966357 Chr.15: 65155812-65155812 on Build GRCh38

View Full Product Details