Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39710023-39710286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB2
Entrez Gene ID
2064
Gene Name
erb-b2 receptor tyrosine kinase 2
Gene Aliases
CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1
UniGene
Hs.446352
Gene Chromosome Location
Chr.17: 39688084-39728662 on Build GRCh38
Transcripts
NM_001005862.2 NM_001289936.1 NM_001289937.1 NM_001289938.1 NM_004448.3 NR_110535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6854238 c.651C>T p.R217R Chr.17: 39710093-39710093 on Build GRCh38
4066010 c.664G>T p.G222C Chr.17: 39710106-39710106 on Build GRCh38
6932253 c.676C>T p.R226C Chr.17: 39710118-39710118 on Build GRCh38
5852395 c.664G>A p.G222S Chr.17: 39710106-39710106 on Build GRCh38
6932254 c.631C>T p.R211C Chr.17: 39710118-39710118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533090552 Chr.17: 39710169-39710169 on Build GRCh38
rs759753872 Chr.17: 39710136-39710136 on Build GRCh38
rs745625627 Chr.17: 39710119-39710119 on Build GRCh38
rs376183465 Chr.17: 39710176-39710176 on Build GRCh38
rs779750598 Chr.17: 39710190-39710190 on Build GRCh38
rs981689859 Chr.17: 39710127-39710127 on Build GRCh38
rs760895559 Chr.17: 39710118-39710118 on Build GRCh38
rs1004581055 Chr.17: 39710097-39710097 on Build GRCh38
rs754564781 Chr.17: 39710236-39710236 on Build GRCh38
rs767309845 Chr.17: 39710170-39710170 on Build GRCh38
rs201102688 Chr.17: 39710239-39710239 on Build GRCh38
rs561177355 Chr.17: 39710051-39710051 on Build GRCh38
rs755873891 Chr.17: 39710177-39710177 on Build GRCh38
rs779346332 Chr.17: 39710092-39710092 on Build GRCh38
rs768151310 Chr.17: 39710091-39710091 on Build GRCh38
rs372694626 Chr.17: 39710050-39710050 on Build GRCh38
rs140636713 Chr.17: 39710117-39710117 on Build GRCh38
rs753474170 Chr.17: 39710202-39710202 on Build GRCh38
rs112495376 Chr.17: 39710203-39710203 on Build GRCh38
rs765424712 Chr.17: 39710159-39710159 on Build GRCh38
rs751504665 Chr.17: 39710057-39710057 on Build GRCh38
rs753984697 Chr.17: 39710049-39710049 on Build GRCh38
rs570351711 Chr.17: 39710047-39710047 on Build GRCh38
rs752656997 Chr.17: 39710163-39710163 on Build GRCh38
rs779058669 Chr.17: 39710090-39710090 on Build GRCh38
rs770976272 Chr.17: 39710089-39710089 on Build GRCh38
rs748701236 Chr.17: 39710105-39710105 on Build GRCh38
rs747570133 Chr.17: 39710248-39710248 on Build GRCh38
rs1040674885 Chr.17: 39710087-39710087 on Build GRCh38
rs772393268 Chr.17: 39710106-39710106 on Build GRCh38
rs776960538 Chr.17: 39710130-39710130 on Build GRCh38
rs747009858 Chr.17: 39710235-39710236 on Build GRCh38
rs373378060 Chr.17: 39710077-39710077 on Build GRCh38
rs757087041 Chr.17: 39710067-39710067 on Build GRCh38
rs755787286 Chr.17: 39710078-39710078 on Build GRCh38

View Full Product Details